Skip to Content
Merck
All Photos(1)

Key Documents

115312

Sigma-Aldrich

N-ω-Methyltryptamine

99%

Synonym(s):

2-(Indol-3-yl)-N-methylethanamine, 3-(2-Methylaminoethyl)indole, 3-(2-[Methylamino]ethyl)indole, N-Monomethyltryptamine, Dipterine, N10-Methyltryptamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2
CAS Number:
Molecular Weight:
174.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

87-89 °C (lit.)

functional group

amine

SMILES string

CNCCc1c[nH]c2ccccc12

InChI

1S/C11H14N2/c1-12-7-6-9-8-13-11-5-3-2-4-10(9)11/h2-5,8,12-13H,6-7H2,1H3

InChI key

NCIKQJBVUNUXLW-UHFFFAOYSA-N

Application

N-ω-Methyltryptamine was used in the preparation of N-acetyl-α−methyltryptamine.
N-ω-methyltryptamine was used in the biosynthesis of dolichantoside using U. tomentosa protein extracts.
Reactant for preparation of:
  • Manzamine analogues for the control of neuroinflammation and cerebral infections
  • Serotonin 4 receptors (5-HT4) receptor agonists
  • A sulful-containing indole alkaloid, glypetelotine
  • Selective inhibitors of cyclin dependent kinase (CDK4)
  • Antagonist of the human tachykinin NK-2 receptor
  • Inhibitors of the tyrosine-specific protein kinase pp60c-src SH2 Domain

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T J Williams et al.
European journal of pharmacology, 245(3), 197-201 (1993-05-15)
The binding of [3H]5-hydroxytryptamine (5-HT) to rat enteric membranes was inhibited by the inclusion of 5-HT 2-methyl-5-HT, 5-hydroxytryptophan, N,N,N-triethyltryptamine and 2-Br-N,N-diethyltryptamine in the incubation buffer. In contrast, tryptamine, 5-methoxytryptamine and 2-methyl-N,N-diethyltryptamine enhanced binding. Ascorbate and dithiothreitol facilitated and reduced binding
R W Walker et al.
Journal of chromatography, 289, 223-229 (1984-04-27)
A capillary column gas-liquid chromatography selected ion monitoring-based method was developed for the measurement of [13C,15N]N-methyltryptamine ( NMT ) in human urine. The method was employed to establish the extent of conversion of [13C,15N]tryptamine to the correspondingly labeled NMT in
Strictosidine-related enzymes involved in the alkaloid biosynthesis of Uncaria tomentosa root cultures grown under oxidative stress.
Vera-Reyes I, Huerta-Heredia AA, Ponce-Noyola T, et al.
Biotechnology Progress, doi:10-doi:10 (2013)
Luigi Servillo et al.
Journal of agricultural and food chemistry, 60(37), 9512-9518 (2012-09-11)
The occurrence of N-methylated tryptamine derivatives in bergamot plant (Citrus bergamia Risso et Poit) is reported for the first time. Interestingly, the most abundant of these substances is N,N,N-trimethyltryptamine, which has not been previously identified in any citrus plant. The
R N Cory et al.
The Journal of pharmacology and experimental therapeutics, 236(1), 48-54 (1986-01-01)
The contractile response of the isolated rabbit aorta elicited by 5-hydroxytryptamine (5-HT) and five partial agonists acting on the 5-HT2 receptor were separated into a phasic and a tonic response by altering the [Ca++] in the buffer. A kinetic analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service