Skip to Content
Merck
All Photos(3)

Key Documents

108448

Sigma-Aldrich

Cyanoacetamide

99%

Synonym(s):

2-Cyanoacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CONH2
CAS Number:
Molecular Weight:
84.08
Beilstein:
878221
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

mp

119-121 °C (lit.)

solubility

cold water: soluble 1gm in 6.5ml

functional group

amide
nitrile

SMILES string

NC(=O)CC#N

InChI

1S/C3H4N2O/c4-2-1-3(5)6/h1H2,(H2,5,6)

InChI key

DGJMPUGMZIKDRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyanoacetamide is the starting reagent for vitamin B6 synthesis[1]. It reacts with reducing carbohydrates in borate buffer to give intense fluorescence andis useful for the automated analysis of carbohydrates as borate complexes[2].

Cyanoacetamide is a highly reactive compound used as a reaction intermediate in condensation and substitution reactions. [3] [4]

Application

Cyanoacetamide was used in spectrofluorimetric method for the determination of some antihistaminic H1 receptor antagonist drugs such as ebastine, cetirizine dihydrochloride and fexofenadine hydrochloride[5]. It may be used in fluorometric determination of 3,4-dihydroxyphenylalanine[6]. It was used as post column fluorigenic derivatizing agent for the bio-analysis and pharmacokinetics of chitosan ester in rabbit serum[5].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

419.0 °F - closed cup

Flash Point(C)

215 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Chen et al.
Scientific reports, 8(1), 13433-13433 (2018-09-09)
Early damage to transplanted organs initiates excess inflammation that can cause ongoing injury, a leading cause for late graft loss. The endothelial glycocalyx modulates immune reactions and chemokine-mediated haptotaxis, potentially driving graft loss. In prior work, conditional deficiency of the
Cyanoacetamide derivatives as synthons in heterocyclic synthesis
Fadda, et al.
Turkish Journal of Chemistry, 32, 259-286 (2008)
Fluorometric determination of 3, 4-dihydroxyphenylalanine with 2-cyanoacetamide.
Liu Y, et al.
Journal of Fluorescence, 13(2), 123-128 (2003)
Fluorimetric determination of reducing carbohydrates with 2-cyanoacetamide and application to automated analysis of carbohydrates as borate complexes.
Honda S, et al.
Analytical Chemistry, 52(7), 1079-1082 (1980)
Cyanoacetamide derivatives as synthons in heterocyclic synthesis.
Fadda AA, et al.
Turkish Journal of Chemistry, 32(3), 259-286 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service