Skip to Content
Merck
All Photos(1)

Documents

690414

Sigma-Aldrich

Malononitrile

Arxada quality, ≥99.0% (calculated, GC, KF)

Synonym(s):

Dicyanomethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(CN)2
CAS Number:
Molecular Weight:
66.06
Beilstein:
773697
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (calculated, GC, KF)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

impurities

≤0.10% water
≤0.50% (E)-2-butenedinitrile
≤0.50% (Z)-2-butenedinitrile
≤0.50% butanedinitrile

bp

220 °C (lit.)

mp

30-32 °C (lit.)

density

1.049 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

N#CCC#N

InChI

1S/C3H2N2/c4-2-1-3-5/h1H2

InChI key

CUONGYYJJVDODC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Malononitrile is an active methylene reagent useful for condensation reactions to synthesize various synthetic intermediates and heterocycles. It is extensively used in the Knoevenagel condensation with various aldehydes and ketones.
Some of the reactions where malononitrile is used as a reactant are:
  • Synthesis of 2-pyran-4-ylidene-malononitrile (PM) based red light emitting polymers.
  • Synthesis of polysubstituted dihydropyridines.
  • Synthesis of various chromene derivatives upon treating with salicylic aldehydes.
  • Synthesis of triselenium dicyanide by treating it with selenium dioxide.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Triselenium dicyanide from malononitrile and selenium dioxide. One-pot synthesis of selenocyanates.
Kachanov, Andrey V et al.
Tetrahedron Letters, 45(23), 4461-4463 (2004)
Synthesis of polysubstituted dihydropyridines by four-component reactions of aromatic aldehydes, malononitrile, arylamines, and acetylenedicarboxylate.
Sun, Jing et al.
Organic Letters, 12(16), 3678-3681 (2010)
Malononitrile.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2001)
Synthesis and characterization of new red-emitting polyfluorene derivatives containing electron-deficient 2-pyran-4-ylidene- malononitrile moieties.
Peng, Qiang et al.
Macromolecules, 37(2), 260-266 (2004)
The condensation of salicylaldehydes and malononitrile revisited: synthesis of new dimeric chromene derivatives.
Costa, Marta et al.
The Journal of Organic Chemistry, 73(5), 1954-1962 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service