Skip to Content
Merck
All Photos(2)

Key Documents

102253

Sigma-Aldrich

2,5-Dimethylaniline

99%

Synonym(s):

2,5-Xylidine, 2-Amino-1,4-dimethylbenzene, 2-Amino-p-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
2205178
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.559 (lit.)

bp

218 °C (lit.)

mp

11.5 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)c(N)c1

InChI

1S/C8H11N/c1-6-3-4-7(2)8(9)5-6/h3-5H,9H2,1-2H3

InChI key

VOWZNBNDMFLQGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Dimethylaniline is a colorless to dark yellow and colorless to brown liquid. It is a substrate for fungal laccases. 2,5-Dimethylaniline undergoes electrochemical oxidation in anhydrous NH4F·2.35 HF medium to form conducting polymer, poly(2,5-dimethylaniline). Poly(2,5-dimethylaniline) coating on copper substrate protects the Cu against corrosion. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied. It induced transcription of lccIV.

Application

2,5-Dimethylaniline was used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation of 2, 5-dimethylaniline in NH 4 F? 2.35 HF medium Characteristics of the resulting conducting polymer.
Genies EM and Noel P.
J. Electroanal. Chem. Interfac. Electrochem., 296(2), 473-490 (1990)
Poly (2, 5-dimethylaniline)-MWNTs nanocomposite: a new material for conductometric acid vapours sensor.
Bavastrello V, et al.
Sensors and Actuators B, Chemical, 98(2), 247-253 (2004)
Luane Ferreira Garcia et al.
Chemosphere, 186, 519-526 (2017-08-16)
The bioremediation and electro-oxidation (EO) processes are included among the most promising cleaning and decontamination mechanisms of water. The efficiency of bioremediation is dictated by the biological actuator for a specific substrate, its suitable immobilization and all involved biochemical concepts.
Thomas Bertrand et al.
Biochemistry, 41(23), 7325-7333 (2002-06-05)
Laccases are multicopper oxidases that catalyze the oxidation of a wide range of phenols or arylamines, and their use in industrial oxidative processes is increasing. We purified from the white rot fungus Trametes versicolor a laccase that exists as five
E Ong et al.
Gene, 196(1-2), 113-119 (1997-10-10)
Laccases are oxidoreductase enzymes involved in the oxidation of various phenolic compounds. They may play a role in the biodegradation of lignin and in the dechlorination of chlorophenols. The cDNAs encoding laccase LccI and a putative laccase LccIV and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service