Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1754

Sigma-Aldrich

Choline bromide

~99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H14NO · Br
CAS Number:
Molecular Weight:
184.07
MDL number:
UNSPSC Code:
12352200

assay

~99%

SMILES string

Br.C[N](C)(C)CCO

Biochem/physiol Actions

Acyl group acceptor

Substrates

Choline acetyltransferase substrate

replaced by

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

188.6 °F - Solvent

flash_point_c

87 °C - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service