Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

585207

Sigma-Aldrich

TG-SDS Buffer, 10X Powder Pack, ULTROL® Grade

Each pack provides an adequate amount of material to make 1 L of 10X concentrate that can be diluted to a 1X solution containing 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3.

Synonym(s):

10X Tris-Glycine-SDS Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

shipped in

ambient

Storage temp.

10-30°C

General description

Each pack provides an adequate amount of material to make 1 L of 10X concentrate that can be diluted to a 1X solution containing 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3.
Each pack provides an adequate amount of material to make 1 L of 10X concentrated buffer that can be diluted to a 1X solution containing 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3. Researchers may check and adjust the final pH using a sensitive pH meter.

Warning

Toxicity: Irritant (B)

Preparation Note

Dissolve one pack in 1 L of water to yield a 10X concentrated solution. The 10X material can be used as 10X, or diluted in the ratio of 1:10 for a 1X solution. Usage criteria should be determined for individual applications.

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at 20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service