Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

492905

Sigma-Aldrich

Nucleozin

A cell-permeable isoxazolylpiperazine compound that is shown to inhibit influenza A H1N1 (A/WSN/33 strain), H3N2 (clinical isolate), and H5N1 (A/Vietnam/1194/04) replication in MDCK cells.

Synonym(s):

Nucleozin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$219.00

$219.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$219.00

About This Item

Empirical Formula (Hill Notation):
C21H19ClN4O4
CAS Number:
Molecular Weight:
426.85
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

$219.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Quality Level

assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellow

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1c(ccc(c1)[N+](=O)[O-])N2CCN(CC2)C(=O)c3c(n[o]c3C)c4ccccc4

InChI

1S/C21H19ClN4O4/c1-14-19(20(23-30-14)15-5-3-2-4-6-15)21(27)25-11-9-24(10-12-25)18-8-7-16(26(28)29)13-17(18)22/h2-8,13H,9-12H2,1H3

InChI key

OWXBJAPOSQSWAO-UHFFFAOYSA-N

General description

A cell-permeable isoxazolylpiperazine compound that is shown to inhibit influenza A H1N1 (A/WSN/33 strain), H3N2 (clinical isolate), and H5N1 (A/Vietnam/1194/04) replication in MDCK cells in vitro (EC50 = 69, 160, and 330 nM, respectively) and significantly reduce fatality after A/Vietnam/1194/04 infection in mice in vivo (50% survial rate on day 21 with 14 b.i.d. i.p. doses of 230 µg/mouse in the first 7 days after infection) by preventing nucleoprotein (NP) nuclear accumulation via NP aggregation induction during early stages of viral infection. Virus strains with N309K or Y289H NP mutation, such as swine-origin influenza A (S-OIV) H1N1, are found to be resistant to Nucleozin, consistent with the N309-mediated hydrogen bonding and Y289-mediated hydrophobic interaction with Nucleozin as predicted by computer-aided docking studies.

Warning

Toxicity: Standard Handling (A)

Other Notes

Kao, R.Y., et al. 2010. Nat. Biotechnol.28, 600.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin E Nilsson-Payant et al.
Journal of virology, 95(9) (2021-02-12)
Negative-sense RNA viruses (NSVs) rely on prepackaged viral RNA-dependent RNA polymerases (RdRp) to replicate and transcribe their viral genomes. Their replication machinery consists of an RdRp bound to viral RNA which is wound around a nucleoprotein (NP) scaffold, forming a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service