Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

382155

Sigma-Aldrich

Histone Deacetylase Substrate, Fluorogenic

Synonym(s):

Histone Deacetylase Substrate, Fluorogenic, Boc-Lys(Ac)-AMC, MAL, N-(4-Methyl-7-coumarinyl)-N α-(t-butoxycarbonyl)-N ω-acetyllysinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31N3O6
CAS Number:
Molecular Weight:
445.51
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 1 mg/mL
DMSO: 5 mg/mL

fluorescence

λex 330 nm
λem 395 nm

shipped in

ambient

storage temp.

−20°C

InChI

1S/C23H31N3O6/c1-14-12-20(28)31-19-13-16(9-10-17(14)19)25-21(29)18(8-6-7-11-24-15(2)27)26-22(30)32-23(3,4)5/h9-10,12-13,18H,6-8,11H2,1-5H3,(H,24,27)(H,25,29)(H,26,30)/t18-/m0/s1

InChI key

SUTRDULVNIPNLW-SFHVURJKSA-N

General description

A fluorogenic substrate for histone deacetylase (HD; excitation maximum: 330 nm; emission maximum: 395 nm).



Note: The histone deacetylase will cleave the Ac group and after the enzymatic reaction 2 peaks will be visible on HPLC. Before the reaction HPLC shows only one peak. The measure of second peak gives the degree of hydrolysis. It is important to use HPLC equipped with a fluorescence detector.

Warning

Toxicity: Standard Handling (A)

Other Notes

Hoffmann, K., et al. 1999. Nucleic Acids Res.27, 2057.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service