371758
GPR81 Agonist, 3Cl-5OH-BA - CAS 53984-36-4 - Calbiochem
Synonym(s):
GPR81 Agonist, 3Cl-5OH-BA - CAS 53984-36-4 - Calbiochem, Hydroxycarboxylic Acid Receptor 1 Agonist, 3-Chloro-5-hydroxybenzoic acid
About This Item
assay
≥98% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
light beige
solubility
DMSO: 100 mg/mL
shipped in
ambient
storage temp.
2-8°C
General description
Biochem/physiol Actions
GPR81
Packaging
Warning
Other Notes
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service