Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239821

Sigma-Aldrich

CXCR4 Antagonist II

The CXCR4 Antagonist II, also referenced under CAS 1258011-83-4, controls the biological activity of CXCR4. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

CXCR4 Antagonist II, 1,3-Dicyclohexyl-2-(3-methyl-6,6-dimethyl-5,6-dihydroimidazo[1,2-b]thiazole)-2-thiopseudourea, HCl, Fusin Antagonist II, IT1t

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34N4S2 · xHCl
CAS Number:
Molecular Weight:
406.65 (free base basis)
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A thiopseudourea compound that acts as a selective and highly potent CXCR4/Fusin antagonist (IC50 = 1.1 nM against SDF-1/CXCL12-induced Ca2+ mobilization in CEM cells) and effectively targets CXCR4-mediated binding of both CXCL12 (IC50 = 8 and 11 nM in competitive binding assays using human CEM and rat IR983F membrane, respectively) and HIV gp120 (IC50 = 7 nM in HIV attachment assays). Shown to be orally available in rat with good in vivo pharmacokinetics (32% availability, tmax = 0.5 h, Cmax= 18 ng/ml, t1/2 = 7.6 h, based on an oral dose of 1 mg/kg). Comparing to AMD3100 (Cat. No. 239820), CXCR4 Antagonist II is ~30-times more potent as a CXCR4-CXCL12 interaction blocker, while being ~2-fold less potent in blocking gp120-mediated HIV attachment.
A thiopseudourea compound that acts as a selective and highly potent CXCR4/Fusin antagonist (IC50 = 1.1 nM against SDF-1/CXCL12-induced Ca2+ mobilization in CEM cells) and effectively targets CXCR4-mediated binding of both CXCL12 (IC50 = 8 and 11 nM in competitive binding assays using human CEM and rat IR983F membrane, respectively) and HIV gp120 (IC50 = 7 nM in HIV attachment assays). Shown to be orally available in rat with good in vivo pharmacokinetics (32% availability, tmax = 0.5 h, Cmax= 18 ng/ml, t1/2 = 7.6 h, based on an oral dose of 1 mg/kg). Comparing to AMD3100 (Cat. No. 239820), CXCR4 Antagonist II is ~30-times more potent as a CXCR4-CXCL12 interaction blocker, while being ~2-fold less potent in blocking gp120-mediated HIV attachment.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wu, B., et al. 2010. Science330, 1066.
Thoma, G., et al. 2008. J. Med. Chem.51, 7915.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service