219511
Cdc2-Like Kinase Inhibitor IV, KH-CB19
The Cdc2-Like Kinase Inhibitor IV, KH-CB19 controls the biological activity of Cdc2-Like KInase. This small molecule/inhibitor is primarily used for Cell Structure applications.
Synonym(s):
Cdc2-Like Kinase Inhibitor IV, KH-CB19, ( E)-Ethyl-3-(2-amino-1-cyanovinyl)-6,7-dichloro-1-methyl-1H-indole-2-carboxylate, Clk Inhibitor IV, (E)-Ethyl-3-(2-amino-1-cyanovinyl)-6,7-dichloro-1-methyl-1H-indole-2-carboxylate, Clk Inhibitor IV
About This Item
Recommended Products
Quality Level
assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 50 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
ClC1=C(Cl)C(N(C)C(C(OCC)=O)=C2/C(C#N)=C\N)=C2C=C1
General description
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service