Biotinylated Peptides
SigmaScreen™ HC Streptavidin Coated Plates, (P9740)
Product No: S6940 (96-well, clear)
Product No: S8815 (384-well, clear)
Our Streptavidin HC (High-Capacity) multiwell plates are prepared with a highly-porous, high-density streptavidin coating. Streptavidin has similar biotin-binding characteristics as avidin. Streptavidin can bind 4 moles of biotin per mole of protein with high selectivity and affinity (Kd~10-15). Unlike avidin, streptavidin has a near neutral pI that alleviates non-specific binding commonly associated with the basic avidin protein.
These plates provide high capacity affinity binding of biotinylated compounds
making them useful for:
- Purification of biotinylated compounds from heterogeneous mixtures.
- Preparation of high capacity affinity matrices for purification of binding partners to immobilized biotinylated compounds.
- Recovery of binding partners and/or biotin compounds for subsequent analysis, e.g. mass spectrometry (MS) and SDS-PAGE.
- Recovery of ICAT™ labeled compounds.
Biotin Binding Capacity | ||
---|---|---|
Product Number | Minimum pmoles Biotin Binding Capacity per Well | |
S 6940 | 96-well | 300 |
S 8815 | 384-well | 150 |
Capacity for biotin based on binding biotin-fluorescein. Steric hindrance will reduce the capacity of the plate when binding larger (> 5 kDa) molecules.
View Protocol Diagram
Data
- Selectivity of Binding on Streptavidin HC
- Comparison of Streptavidin HC Plate to Monomeric Avidin Agarose