MilliporeSigma
HomeqPCRChoosing the Right Custom qPCR Probe
Choosing the Right Custom qPCR Probe
There is a wide selection of possible custom probes that can be used for qPCR, and each has advantages for different applications. The summary below highlights the applications of commonly used custom qPCR probes. 
Application Reference Guide
 SYBR® Green I*Dual-Labeled ProbesDual-Labeled
Locked Nucleic Acid
Probes
Molecular Beacons*LightCycler® Probes*
APPLICATION
Mass screening• •    
Microarray validation• •   
Multiple target genes / few samples   
SNP detection  • •
Allele discrimination  • •
Pathogen detection• •
Multiplexing • •• •• •
Viral load quantification • •
Gene expression analysis
• •• •• •• •
Gene copy determination • •
Endpoint genotyping   • • 
in vitro quantification or detection  • • 
Table 1.*Locked Nucleic Acid can be incorporated into the PCR Primers or qPCR probes for improved specificity. Locked Nucleic Acid in oligonucleotides other than qPCR probes is available in all countries except for the United States.
Reporters, Quenchers, and Instrument Compatibility
Modern qPCR platforms typically have multiple detection channels enabling flexibility in the choice of probe labels. It is important to select the fluorescent labels which are compatible with the detection channels for the qPCR instrument and to ensure the correct filter settings or detection calibration for the instrument. The Reporters and Instrument Compatibility table lists a selection of some of the most widely used qPCR platforms and indicates which fluorescent labels may be used. Please note that not all labels are listed and many alternative reporters are available. For information on the use of non-standard labels with these platforms, contact your local technical service professional. 
Dye Substitutes
Several qPCR instruments utilize proprietary dyes which are not generally available commercially, such as VIC® and NED™. When seeking dye alternatives, the following criteria are important:
  • The excitation and detection wavelength are compatible with the instrument light source and detection system
  • For probes, the quencher effectively absorbs light at the emission wavelength of the reporter
  • The higher the extinction coefficient the brighter the dye, which contributes to sensitive detection 
  • When using multiple dyes (multiplex) the excitation and emission wavelengths of each dye must be independent to avoid cross talk
Quenchers
Quenching molecules are typically placed at the 3' end of single molecule probes such as Dual-Labeled Probes and Molecular Beacons. Quenchers may be fluorescent (TAMRA™) or nonfluorescent molecules (DABCYL, Black Hole Quencher™ (BHQ™). For optimal performance, the quencher's absorbance spectrum should match the reporter’s emission spectrum as closely as possible. Recommended reporter/quencher combinations can be found in the Spectral Properties table.
Reporters, Quenchers, and Instrument Compatibility
Modern qPCR platforms typically have multiple detection channels enabling flexibility in the choice of probe labels. It is important to select the fluorescent labels which are compatible with the detection channels for the qPCR instrument and to ensure the correct filter settings or detection calibration for the instrument. The Reporters and Instrument Compatibility table lists a selection of some of the most widely used qPCR platforms and indicates which fluorescent labels may be used. Please note that not all labels are listed and many alternative reporters are available. For information on the use of non-standard labels with these platforms, contact your local technical service professional. 
Dye Substitutes
Several qPCR instruments utilize proprietary dyes which are not generally available commercially, such as VIC® and NED™. When seeking dye alternatives, the following criteria are important:
  • The excitation and detection wavelength are compatible with the instrument light source and detection system
  • For probes, the quencher effectively absorbs light at the emission wavelength of the reporter
  • The higher the extinction coefficient the brighter the dye, which contributes to sensitive detection 
  • When using multiple dyes (multiplex) the excitation and emission wavelengths of each dye must be independent to avoid cross talk
Quenchers
Quenching molecules are typically placed at the 3' end of single molecule probes such as Dual-Labeled Probes and Molecular Beacons. Quenchers may be fluorescent (TAMRA™) or nonfluorescent molecules (DABCYL, Black Hole Quencher™ (BHQ™). For optimal performance, the quencher's absorbance spectrum should match the reporter’s emission spectrum as closely as possible. Recommended reporter/quencher combinations can be found in the Spectral Properties table.
DyeMax.
EX (nm)
Max.
EM (nm)
Compatible
Quencher
6-FAM™495520BHQ™-1, TAMRA
JOE™529555BHQ™-1, TAMRA
TET™521536BHQ™-1, TAMRA
Cal Fluor® Gold 5401522544BHQ™-1
HEX™2535556BHQ™-1, TAMRA
Cal Fluor Orange 5602538559BHQ™-1
TAMRA557583BHQ™-2
Cyanine 3549566BHQ™-2
Quasar® 5703548566BHQ™-2
Cal Fluor Red 5904569591BHQ™-2
ROX™586610BHQ™-2
TxRd (Sulforhodamine 101-X)597616BHQ™-2
Cyanine 5646669BHQ™-3
Quasar 6705647670BHQ™-3
Cyanine 5.5675694BHQ™-3
1JOE/TET alternative
2VIC® alternative
3Cyanine 3 alternative
4TAMRA alternative
5Cyanine 5 alternative
Table 2.Spectral Properties Table
PlatformSYBR
Green I
FAMHEXJOEROXTETCya-
nine 3
Cya-
nine 5
TAMRATxRdLC Red
640
LC Red
705
ABI 7900HT     
ABI 7300      
ABI 7500   
ABI 7700      
ABI 7000       
ABI StepOne™     
ABI StepOnePlus™     
Bio-Rad iQ™ 5
Bio-Rad Opticon® 2       
Bio-Rad Chromo4™  
Bio-Rad MyiQ™
Bio-Rad MiniOpticon™
Bio-Rad CFX96™
Bio-Rad SFX384™
Agilent Mx4000®  
Agilent Mx3000P®  
Agilent Mx3005P®  
Roche LightCycler®        
Roche LightCycler 2       
Roche LightCycler 480      
Cepheid SmartCycler®       
Cepheid SmartCycler II      
Qiagen Rotor-Gene® 6000   
Eppendorf Mastercycler® realplex     
Note: Not all qPCR instruments or fluorophores are listed. Contact the instrument manufacturer for details on compatible fluorophores
Table 3.Reporters and Instrument Compatibility Table