Counting of Cells
Trypan blue method of cell counting is inexpensive and readily available method. The method is based on principle that living cells exclude trypan blue dye and appear bright, while dead cells take up dye and appear blue. Watch video to know the procedure of trypan blue method of cell counting.