Basal Media Formulation
Component | B1522 [1x] g/L | B9638 g/L |
---|---|---|
Inorganic Salts | ||
Calcium Chloride | 0.2 | 0.2 |
Magnesium Sulfate (anhydrous) | 0.09767 | 0.09767 |
Potassium Chloride | 0.4 | 0.4 |
Sodium Bicarbonate | 2.2 | — |
Sodium Chloride | 6.8 | 6.8 |
Sodium Phosphate Monobasic (anhydrous) | 0.122 | 0.122 |
Amino Acids | ||
L-Arginine • HCl | 0.021 | 0.021 |
L-Cystine • 2HCl | 0.01565 | 0.01565 |
L-Glutamine | — | 0.292 |
L-Histidine (free base) | 0.008 | 0.008 |
L-Isoleucine | 0.026 | 0.026 |
L-Leucine | 0.026 | 0.026 |
L-Lysine • HCl | 0.03647 | 0.03647 |
L-Methionine | 0.0075 | 0.0075 |
L-Phenylalanine | 0.0165 | 0.0165 |
L-Threonine | 0.024 | 0.024 |
L-Tryptophan | 0.004 | 0.004 |
L-Tyrosine • 2Na • 2H2O | 0.02595 | 0.02595 |
L-Valine | 0.0235 | 0.0235 |
Vitamins | ||
D-Biotin | 0.001 | 0.001 |
Choline Chloride | 0.001 | 0.001 |
Folic Acid | 0.001 | 0.001 |
myo-Inositol | 0.002 | 0.002 |
Niacinamide | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 | 0.001 |
Pyridoxal • HCl | 0.001 | 0.001 |
Riboflavin | 0.0001 | 0.0001 |
Thiamine • HCl | 0.001 | 0.001 |
Other | ||
D-Glucose | 1 | 1 |
Phenol Red • Na | 0.011 | 0.011 |
Add | ||
L-Glutamine | 0.292 | — |
Sodium Bicarbonate | — | 2.2 |