MilliporeSigma
HomeEnzyme Activity AssaysEnzymatic Assay of Pepsin (3.4.23.1)
Enzymatic Assay of Pepsin (3.4.23.1)
Description
This procedure may be used for determination of Pepsin activity using hemoglobin as the substrate. It is a spectrophotometric stop rate determination.
Unit Definition: One unit of Pepsin will produce a ΔA280 of 0.001 per minute at pH 2.0 at 37 °C measured as trichloroacetic acid (TCA)-soluble products using hemoglobin as the substrate. (Final volume = 16 ml, light path = 1 cm.)
Reagents and Equipment Required
1.0 M Hydrochloric Acid
Hemoglobin from bovine blood (Catalog Number H2625)
6.1 N [~100% (w/v)] Trichloroacetic acid solution (Catalog Number T0699)
Precautions
Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Preparation Instructions
Use ultrapure water (≥18 MΩxcm resistivity at 25 °C) for the preparation of reagents.
10 mM HCl (Hydrochloric Acid) – Dilute 1.0 M Hydrochloric Acid 100‑fold with ultrapure water.
2.5% (w/v) Hemoglobin Stock Solution – Prepare a 25 mg/ml solution in ultrapure water using Hemoglobin from bovine blood (Catalog Number H2625). Stir vigorously, creating a vortex, for a minimum of 10 minutes at 37 °C. Then filter through a polypropylene column with a coarse filter (90–130 mm).
Substrate [2.0% (w/v) Hemoglobin solution] – Transfer 80 ml of the filtered 2.5% (w/v) Hemoglobin Stock Solution into a suitable container. Adjust the pH of this solution to 2.0 at 37 °C using 5 M HCl. Bring the final volume to 100 ml with ultrapure water.
TCA Solution [5% (w/v) Trichloroacetic Acid] – Dilute 6.1 N [~100% (w/v)] Trichloroacetic acid solution (Catalog Number T0699) 20-fold with ultrapure water.
Enzyme Solution (Pepsin) – Prepare a 1 mg/mL stock solution in cold (2–8 °C) 10 mM HCl. If insoluble material is present, allow the stock solution to sit on ice until dissolved. When the Pepsin has dissolved or, if insoluble material is still present, after 1 hour, dilute the 1 mg/mL stock solution further to 0.01–0.05 mg/ml Enzyme Solution in cold 10 mM HCl.
Procedure
1. Pipette the Substrate into suitable glass vials.
ReagentBlank (ml)Test 1 (ml)Test 2 (ml)Test 3 (ml)
Substrate5.005.005.005.00
2. Place vials in a thermostatted water bath and equilibrate to 37 °C for ~10 minutes, then add:
ReagentBlank (ml)Test 1 (ml)Test 2 (ml)Test 3 (ml)
Enzyme Solution1.001.001.00
3. Mix by swirling and incubate at 37 °C for exactly 10 minutes, then add:
ReagentBlank (ml)Test 1 (ml)Test 2 (ml)Test 3 (ml)
TCA Solution10.010.010.010.0
Enzyme Solution1.00
4. Mix by swirling and incubate at 37 °C for an additional 5 minutes.
5. Filter the Blank and Test mixtures through a 0.45 µm syringe filter. Using a spectrophotometer, record the A280 versus air for each filtered solution for each vial.
Units/ml enzyme =
(A280 Test – A280 Blank) x (df)
(10) x (1.0) x (.001)
where:
df = Dilution factor
10 = Assay incubation time in minutes
1.0 = Volume of Enzyme Solution (ml) added
0.001 = ΔA280 per unit of Pepsin (unit definition)
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Reference
1.
Anson ML. 1938. THE ESTIMATION OF PEPSIN, TRYPSIN, PAPAIN, AND CATHEPSIN WITH HEMOGLOBIN. 22(1):79-89. https://doi.org/10.1085/jgp.22.1.79