MilliporeSigma
HomeApplicationsMaterials Science and Engineering Microelectronics & Nanoelectronics
Microelectronics & Nanoelectronics
Illustration of printed circuit board made of electronic components at the micrometer and nanometer scale.
Microelectronics and nanoelectronics are subfields of electronics in which the nominal feature sizes of electronic components are between 100 and 0.1 micrometers in magnitude (microelectronics) or 100 nanometers or smaller (nanoelectronics). The memory storage power of today’s advanced electronic devices has been achieved by significantly increasing the density of microchips. By decreasing the size of field-effect transistors, more components can be fit into integrated circuits, allowing for more powerful and energy-efficient electronic devices with reduced weights and power consumption.
According to Moore’s Law, the number of transistors that can be put on a single chip will double every two years. Since this was projected in 1965, semiconductor fabrication technology sustained this rate of advancement and revolutionized the industry. However, pace of dimension reduction is slowing, and the key challenge in fabricating electronic components in the sub-micrometer range is the design of the transistor gate, which controls the current flow in the channel. The smaller electronic components are, the more challenging they become to manufacture. Physical and quantum effects alter materials’ properties from a macroscale to a nanoscale, influencing inter-atomic interactions and quantum mechanical properties.
Related Technical Articles
  • Graphene has emerged as the new wonder material. Being only one atom thick and composed of carbon atoms arranged in a hexagonal honeycomb lattice structure, the interest in this material has exploded exponentially since 2004 when it was first isolated and identified using a very simple method.
  • Find unique properties & applications of single (SWNTs) , double (DWNTs) & multi walled carbon nanotubes (MWCNTs).
  • Graphene oxide is a unique material that can be viewed as a single monomolecular layer of graphite with various oxygen containing functionalities such as epoxide, carbonyl, carboxyl and hydroxyl groups.
  • Thermoelectric materials comprise a wide range of solid compounds distinguished by their ability to convert thermal and electrical energy.
  • Spintronics, short for spin-based electronics, sometimes called magnetoelectronics, is the term given to microelectronic devices that function by exploiting the spin of electrons.
  • See All (64)
Related Protocols
  • Microparticles protocol for washing particles may be done via centrifugation. This procedure must be performed carefully.
  • Our photoresist kit was designed to have the necessary chemical components for each step in the lithographic process. The component materials are provided in pre-weighed quantities for your convenience. Etchants are available separately so that the proper etchant can be chosen for a variety of substrate choices.
  • The dispersibility and bundle defoliation of single-walled carbon nanotubes (SWCNTs), which can be applied to materials produced by the CoMoCAT® process, have been extensively investigated by SouthWest NanoTechnologies (SWeNT ®) and at the University of Oklahoma.
  • Selenium is an essential trace element. It is a necessary dietary constituent of at least 25 human selenoproteins and enzymes containing selenocysteine. Additionally, as selenium is a semiconductor and photoelectrically active, it has more advanced applications such as xerography and solar cell assembly.
  • The following material related to Nanodisc Technology is adapted from on-line content of the research group of Professor Stephen Sligar of the University of Illinois at Urbana-Champaign, with the kind permission of Professor Sligar.
  • See All (4)
Find More Articles and Protocols
The arrival of innovative materials, carbon nanotubes, boron nitride nanotubes, quantum dots, and graphene additives, have advanced the minimization of nanotechnology and microtechnology. These and other new materials can be shaped and manipulated with extraordinary precision at the tiniest of scales. Novel technologies enable the deposition and layering of electronic materials with precise thickness, even down to the atomic level. Thin-film semiconductor device fabrication technology uses conducting, semiconducting, and insulating materials to deliver advanced capabilities at high volumes and very low cost. Modern manufacturing methods for nanoelectronics include patterning (lithography), etching, thin film deposition, and doping techniques.
Emerging research fields focus on new approaches in nanotechnology and quantum mechanical effects. Molecular electronics uses single molecules as electronic components to establish electrical contact with bulk-sized electrodes. Spintronics, or spin-transport electronics, manipulates electrons’ spin property with magnetic and electric fields, resulting in a spin-polarized current that provides higher data transfer speeds and greater storage capacity, memory density and processing power than is possible with electric charge alone.