MilliporeSigma
HomeCell Counting & Health AnalysisScepter™ 3.0 Handheld Cell Counter
Scepter™ 3.0 Handheld Cell Counter
The Scepter™ 3.0 Cell Counter is the only automated cell counter that's portable and handheld. This cell counter is the only device to bring the accuracy and consistency of Coulter impedance counting right to the culture hood. While other automated counters consume bench space and rely on object recognition software, manual focusing, and clumsy loading chambers, the Scepter™ 3.0 cell counter provides true automation without the risk of error that accompanies vision-based systems. With its microfabricated, precision-engineered sensor, the Scepter™ 3.0 cell counter does all the work of assessment and calculation, and delivers accurate and reliable cell counts in less than 30 seconds.
How it Works / Performance
How it Works / Performance
Besides being the only handheld automated cell counter, the Scepter™ 3.0 counter brings Coulter-based accuracy that visual counters can't provide. Find out how it works, and see the performance data.
Validated Cell Types
Validated Cell Types
The Scepter™ counter has been shown to be accurate over a range of cell types and sizes. See a list of which mammalian and other key phenotypes have been validated for Scepter™ counting precision.
Register Your Scepter™ 3.0
Register Your Scepter™ 3.0
Get your instrument registered with us to ensure you receive updates to software and firmware. Plus, be the first to get timely alerts about new applications, and more.
For Scepter™ 2.0 Users
For Scepter™ 2.0 Users
Already counting smarter with the Scepter™ 2.0 device? Find out here how to upgrade to the more robust, more connected, easier to read version 3.0.