MilliporeSigma
Lateral Flow Membranes
Hi-Flow™ Plus membranes are the membrane of choice for diagnostic tests and kits, offering superior consistency and performance. Uniquely optimized for lateral flow assays, Hi-Flow™ Plus membranes are manufactured to the tightest specifications compared to other lateral flow membranes on the market and offer unsurpassed surface quality. Hi-Flow™ Plus membranes are specifically designed for consistent results, reliable lot-to-lot performance, fixed test run times, and sharp capture / test lines with minimal nonspecific background signal. Hi-Flow™ Plus membranes are suitable for all lateral flow test types and are available in several convenient speeds and formats.
Cut away schematic drawing of a rapid lateral flow assay test strip
Selecting the Right Raw Materials for Your Lateral Flow Test
The membrane is the single most important material used in a lateral flow test strip. Physical and chemical attributes of the membrane affect its capillary flow properties. The capillary flow properties, in turn, affect reagent deposition, assay sensitivity, assay specificity, and test line consistency. Other physical properties of the membrane affect integration into finished test strips. Learn more in our guide, "Rapid Lateral Flow Test Strips - Considerations for Product Development", which provides a deep dive into the key aspects of lateral flow test strip design, particularly the materials used and their integration with reagents and manufacturing processes. Understanding these principles will help provide a framework for rapid lateral flow test development.
Hi-Flow™ Plus Membrane Speeds
The speed at which a sample material moves along a membrane strip is linked to the capillary flow rate of the membrane. Moreover, there is a direct correlation between the speed and sensitivity of the membrane. Understanding these requirements will enable you to select the appropriate membrane for your application. For example, you can use fast-flowing Hi-Flow™ Plus 75 membranes for tests where speed is critical, sample is plentiful, and sensitivity is not a concern. Slow-flowing Hi-Flow™ Plus 180 membranes can be used for tests where speed is less important, sample is limited, or high sensitivity is needed.
Hi-Flow™ Plus Membrane Speeds
MembraneCapillary Flow Time Specification** (sec/4 cm)Flow RateSensitivity
HF180180 ± 45SlowestMost sensitive
HF135135 ± 34
HF120120 ± 30
HF09090 ± 23
HF07575 ± 19FastestLeast sensitive
Products
Sort by Relevance
Showing 1-20 of 72
Change View
Showing 1-20 of 72
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 72 results
Hi-Flow™ Plus Membrane Formats
Hi-Flow™ Plus Membrane Formats for Lateral Flow Assays products on yellow background.
Hi-Flow™ Plus membranes are available in several formats, from sample sheets and slit rolls for evaluation purposes to bulk master rolls slit to desired widths for manufacturing purposes. These membranes are also offered with different backing options depending on individual assay requirements. Preassembled membrane cards are also offered, providing an efficient and convenient choice for manufacturers by eliminating the need to manually assemble cards for development or production of rapid diagnostic kits and tests.
Hi-Flow™ Plus Variety Sample Sheet Packs
Convenient sample packs offer a variety of sheets from the Hi-Flow™ Plus membrane portfolio. Ideal for membrane selection and evaluation purposes during the early phase of a lateral flow test development project.
Variety PackBackingDimensionsQuantityProduct Number
Hi-Flow™ Plus 75, 90, 120, 135, 1802 mil25 cm x 30.5 cm sheets10 sheets/pack (2 sheets of each speed)HFSAMPLE02
Hi-Flow™ Plus 75, 90, 120, 135, 1804 mil25 cm x 30.5 cm sheets10 sheets/pack (2 sheets of each speed)HFSAMPLE04
Hi-Flow™ Plus Sample Sheet Packs on yellow background.
Hi-Flow™ Plus Sample Sheet Packs
Sample packs containing five sheets of Hi-Flow™ Plus membrane. Ideal for early phase lateral flow test development.
Product NameBackingDimensionsQuantity/PackProduct Number
Hi-Flow™ Plus 75 Membrane2 mil25 cm x 30.5 cm sheets5HF07502XSS
Hi-Flow™ Plus 75 Membrane4 mil25 cm x 30.5 cm sheets5HF07504XSS
Hi-Flow™ Plus 90 Membrane2 mil25 cm x 30.5 cm sheets5HF09002XSS
Hi-Flow™ Plus 90 Membrane4 mil25 cm x 30.5 cm sheets5HF09004XSS
Hi-Flow™ Plus 120 Membrane2 mil25 cm x 30.5 cm sheets5HF12002XSS
Hi-Flow™ Plus 120 Membrane4 mil25 cm x 30.5 cm sheets5HF12004XSS
Hi-Flow™ Plus 135 Membrane2 mil25 cm x 30.5 cm sheets5HF13502XSS
Hi-Flow™ Plus 135 Membrane4 mil25 cm x 30.5 cm sheets5HF13504XSS
Hi-Flow™ Plus 180 MembraneUB25 cm x 30.5 cm sheets5HF180UBXSS
Hi-Flow™ Plus 180 Membrane2 mil25 cm x 30.5 cm sheets5HF18002XSS
Hi-Flow™ Plus 180 Membrane4 mil25 cm x 30.5 cm sheets5HF18004XSS
Hi-Flow™ Plus Sample Slit Rolls for Lateral Flow Assays on yellow background.
Hi-Flow™ Plus Sample Slit Rolls
Individual sample slit rolls of Hi-Flow™ Plus membrane. These rolls are useful in the latter stages of lateral flow test development once test design has been established and scale-up trials and process optimization are to be commenced.
Product NameBackingDimensionsQuantity/RollProduct Number
Hi-Flow™ Plus 75 Membrane2 mil25 mm width roll100 mSHF0750225
Hi-Flow™ Plus 75 Membrane4 mil25 mm width roll100 mSHF0750425
Hi-Flow™ Plus 90 Membrane2 mil25 mm width roll100 mSHF0900225
Hi-Flow™ Plus 90 Membrane4 mil25 mm width roll100 mSHF0900425
Hi-Flow™ Plus 120 Membrane2 mil25 mm width roll100 mSHF1200225
Hi-Flow™ Plus 120 Membrane4 mil25 mm width roll100 mSHF1200425
Hi-Flow™ Plus 135 Membrane2 mil25 mm width roll100 mSHF1350225
Hi-Flow™ Plus 135 Membrane4 mil25 mm width roll100 mSHF1350425
Hi-Flow™ Plus 180 Membrane2 mil25 mm width roll100 mSHF1800225
Hi-Flow™ Plus 180 Membrane4 mil25 mm width roll100 mSHF1800425
Hi-Flow™ Plus 180 MembraneUB25 mm width roll100 mSHF180UB25
Hi-Flow™ Plus Membrane Master Rolls for Lateral Flow Assay Manufacturing on yellow background.
Hi-Flow™ Plus Membranes for Manufacturing
Hi-Flow™ Plus membranes for manufacturing scale purposes are now available to purchase online in convenient pack formats.
20mm wide Hi-Flow™ Plus slit rolls in a variety of capillary flow speeds and 2 backing options are offered in packs of 14.
25mm wide Hi-Flow™ Plus slit rolls in a variety of capillary flow speeds and 2 backing options are offered in packs of 11.
Available to ship immediately, get the same superior consistency and high-performance membrane, quickly and without the wait.
Product NameBackingDimensionsQuantity/RollProduct Number
Hi-Flow™ Plus Membrane 752 milwidth 20 mmpack of 14 rollBPCKHF07502S20
Hi-Flow™ Plus Membrane 752 milslit width 25 mmpack of 11 rollBPCKHF07502S25
Hi-Flow™ Plus Membrane 754 milslit width 20 mmpack of 14 rollBPCKHF07504S20
Hi-Flow™ Plus Membrane 754 milslit width 25 mmpack of 11 rollBPCKHF07504S25
Hi-Flow™ Plus Membrane 902 milslit width 20 mmpack of 14 rollBPCKHF09002S20
Hi-Flow™ Plus Membrane 902 milslit width 25 mmpack of 11 rollBPCKHF09002S25
Hi-Flow™ Plus Membrane 904 milslit width 20 mmpack of 14 rollBPCKHF09004S20
Hi-Flow™ Plus Membrane 904 milslit width 25 mmpack of 11 rollBPCKHF09004S25
Hi-Flow™ Plus Membrane 1202 milslit width 20 mmpack of 14 rollBPCKHF12002S20
Hi-Flow™ Plus Membrane 1202 milslit width 25 mmpack of 11 rollBPCKHF12002S25
Hi-Flow™ Plus Membrane 1204 milslit width 20 mm pack of 14 rollBPCKHF12004S20
Hi-Flow™ Plus Membrane 1204 milslit width 25 mm pack of 11 rollBPCKHF12004S25
Hi-Flow™ Plus Membrane 1352 milslit width 20 mm pack of 14 rollBPCKHF13502S20
Hi-Flow™ Plus Membrane 1352 milslit width 25 mm pack of 11 rollBPCKHF13502S25
Hi-Flow™ Plus Membrane 1354 milslit width 20 mm pack of 14 rollBPCKHF13504S20
Hi-Flow™ Plus Membrane 1354 milslit width 25 mm pack of 11 rollBPCKHF13504S25
Hi-Flow™ Plus Membrane 1802 milslit width 20 mm pack of 14 rollBPCKHF18002S20
Hi-Flow™ Plus Membrane 1802 milslit width 25 mm pack of 11 rollBPCKHF18002S25
Hi-Flow™ Plus Membrane 1804 milslit width 20 mm pack of 14 rollBPCKHF18004S20
Hi-Flow™ Plus Membrane 1804 milslit width 25 mm pack of 11 rollBPCKHF18004S25
Standard (20mm & 25mm) and customized slit widths of Hi-Flow™ Plus membranes are also available to order offline as master rolls. Each master roll is 30.5 cm wide and 100 m in length. For ease of use, master rolls are cut into multiple ready-to-use slit rolls at the desired width prior to delivery. The number of slit rolls received per master roll order is tied to the required width of the slit roll. As an example, an order for 1 master roll with a required slit width of 18mm will yield 16 slit rolls.
Please contact your local account manager or our customer service team to obtain a catalog number for your membrane and slit width of choice.
Hi-Flow™ Plus Membranes for Manufacturing - Slit Roll Widths
Slit Width (cm)Slit Rolls/Master RollLinear Meters/100m Master RollLinear Feet/100m Master Roll
1.61818005904
1.71717005576
1.81616005248
21414004592
2.21313004264
2.251313004264
2.51111003608
2.54 (1 in)1111003608
2.81010003280
399002952
3.17599002952
3.299002952
3.588002625
3.81 (1.5 in)77002297
477002297
4.566001969
4.866001969
555001640
83300984
13.22200656
30.51100238
Hi-Flow™ Plus Membrane Cards for lateral flow assays on yellow background.
Hi-Flow™ Plus Membrane Cards
Hi-Flow™ Plus membrane cards provide an easy-to-use subassembly so you don’t have to allocate time and resources assembling cards, thereby reducing and simplifying your manufacturing design process and production. Utilizing our quality Hi-Flow™ Plus membranes, these cards are assembled in a highly precise manner to deliver consistent performance in your test.
Hi-Flow™ Plus membrane card dimensions
Standard Hi-Flow™ Plus membrane card dimensions
Note: custom membrane card dimensions are also possible.
Product NameCard DimensionsQuantity Cards/PackProduct Number
Millipore Laminated Cards with release liners; No Membrane6 cm x 30 cm100HF000MC100
HI-Flow™ Plus HF075 Membrane Cards6 cm x 30 cm100HF075MC100
HI-Flow™ Plus HF090 Membrane Cards6 cm x 30 cm100HF090MC100
HI-Flow™ Plus HF120 Membrane Cards6 cm x 30 cm100HF120MC100
HI-Flow™ Plus HF135 Membrane Cards6 cm x 30 cm100HF135MC100
HI-Flow™ Plus HF180 Membrane Cards6 cm x 30 cm100HF180MC100
HI-Flow™ Plus HF075 Membrane Cards6 cm x 30 cm5HF075MC5PK
HI-Flow™ Plus HF090 Membrane Cards6 cm x 30 cm5HF090MC5PK
HI-Flow™ Plus HF120 Membrane Cards6 cm x 30 cm5HF120MC5PK
HI-Flow™ Plus HF135 Membrane Cards6 cm x 30 cm5HF135MC5PK
HI-Flow™ Plus HF180 Membrane Cards6 cm x 30 cm5HF180MC5PK
SureWick® Pad Materials for Lateral Flow Assays
SureWick® Pad Materials are used in lateral flow assays as sample, absorbent, and conjugate pads. The 100% pure cellulose fiber sample and absorbent pads have no binders or glues to interfere with assay performance. Glass fiber conjugate pads have low extractables and excellent consistency.
These products are available in a variety of dimensions and formats (sheets, strips, and rolls) to fit your unique application. We carry common sizes in inventory and can customize these products to meet the dimensional requirements of your device and manufacturing process.
DescriptionFormatDimensionsQuantity/PackProduct Number
GFDX Glass Fiber Diagnostic PadsStrips10 mm x 300 mm100GFDX103000
8 mm x 300 mm100GFDX083000
Sheets20 cm x 30 cm5GFDX20305PK
20 cm x 30 cm100GFDX203000
20.32 cm x 25.4 cm100GFDX202500
Rolls8 mm x 50 m1GFDX000850
8 mm x 100 m1GFDX000800
10 mm x 50 m1GFDX001050
10 mm x 100 m1GFDX001000
C083 Cellulose Absorbent Sample PadsStrips17 mm x 300 mm100CFSP173000
20 mm x 300 mm100CFSP203000
Sheets20 cm x 30 cm5CFSP22305PK
20 cm x 30 cm100CFSP223000
Rolls17 mm x 100 m1CFSP001700
20 mm x 100 m1CFSP002000
Hi-Flow™ Plus Membranes and SureWick® Conjugate, Sample, and Absorbent Pads provide a complete solution for your lateral flow assays.
Lateral Flow Assay Development Services
Get fast, expert support on your IVD assay development when you need it. Discover our Assay Development Services for fast-access expert guidance and global lab support for your LFA development or optimization at any stage.
Our new Assay Development Services has:
  • dedicated, fully equipped state-of-the-art labs
  • globally-distributed labs and personnel for fast regional response
  • personnel specifically trained to support any part of LFA development
  • broad, comprehensive knowledge of assay optimization
  • a long history of developing & manufacturing many of the raw materials and key components used in IVDs today
Related Resources
Page 1 of 2
External Resources
The Hidden Manufacturers of COVID-19 Tests Podcast
In this episode, Shawn Gaskell discusses capillary flow membranes used in Covid...
RSV Cases are Rising Fast in Children. Early Detection Can Help.
RSV hospitalizations for 2022 have surged, especially among young children. 