Authenticated Liver Cancer Cell Lines for Cancer Research
Amongst liver cancers, the incidence of hepatocellular carcinoma (HCC) is increasing and has a high mortality rate. Chronic hepatitis B- or hepatitis C-induced cirrhosis is the leading risk factor for HCC. For reasons that are not entirely known, males may be three to five times more likely than females to develop HCC, the most common type of liver cancer. 83% of cases are diagnosed in less-developed countries, with the highest rates found in east and southeast Asia and in middle and western Africa; rates are lower in south-central and western Asia. The five-year survival rate is 31% for patients who are diagnosed at an early stage, but drops to 3% if cancer has metastasized to other tissues of the body.1
Types of Liver Cancer
Hepatocellular carcinoma accounts for 80% of all adult liver cancers. Cholangiocarcinoma (bile duct cancer) and angiosarcoma comprise 10-20% and 1% of liver cancer diagnoses, respectively.
Type of cancer | Hepatocellular carcinoma | Cholangiocarcinoma | Angiosarcoma |
Tissue of origin | Liver | Bile duct tissue | Hepatic blood vessels |
Risk Factors
The major risk factor for liver cancer is exposure to hepatitis viruses and environmental pathogens. Hepatitis B virus (HBV) infection accounts for 60% of total liver cancer, and hepatitis C virus (HCV) is responsible for 33%. Other risk factors include alcohol-related cirrhosis, nonalcoholic fatty liver disease, and obesity.
Mutations
The most commonly mutated genes in liver cancer are TP53, CTNNB1, TERT, HNF1A, LRP1B, ARID1A, AXIN1, ARID2, KMT2C, and IL6ST.
Click on the genes (above) to find relevant products (antibodies, shRNA, siRNA, primers, CRISPR plasmids) for your research study.
Small Molecules/Monoclonal Antibodies
Small molecule compounds and antibodies can be used to target cancer cells and block tumor growth and progression. The most common strategy for hepatocellular carcinoma is inhibition of angiogenesis signals. Drugs used to target liver cancer include:
- Sorafenib Tosylate (Nexavar)
- Regorafenib (Stivarga)
Applications
Cancer cell lines are essential for cancer research, and provide an accessible, cost-effective model for cellular behavior and response. Based on cell phenotype and experimental need, cell lines have the potential for utility in multiple applications. Some examples of application-specific cell line use are included below.
Application | Cell line used |
---|---|
In vitro screening | Hepatic cancer cell lines like Huh-7D12 were used to test the cytotoxic effects of plant extracts3 and chemical compounds4 |
Target identification/validation | Hepatocellular carcinoma cell lines, including HepG2, Hep3B, Huh7, C3A, PLC, LO2, and SUN387 were used to validate the role of p53-miR-1246-NFIB pathway in disease pathogenesis5 |
Toxicology studies | Spheroids derived from HepG2 were used for repeated dose high-throughput toxicity studies6 |
Metastasis studies | SK-HEP-1 cells were used to screen drugs that inhibit metastasis7 |
Xenograft models | HepG2 cell lines were used to develop xenograft models of hepatocellular carcinoma8 |
In-vitro models | HepG2 cells were modeled to study the life cycle of hepatitis B virus9 Rat hepatocytes (McA RH-7777) and human hepatocytes (HepG2) were used as in vitro models for ischemia/reperfusion diseases10 |
miRNA regulation studies | NCTC 1469 cell lines were used to study the role of MiR-19a in glycogen synthesis of hepatocytes11 |
ECACC Liver Cancer Cell Lines | ||
---|---|---|
Product No. | Cell Name | Cell Line Origin |
87050701 | ARL-6 | Rat Wistar liver hepatoma |
94101906 | C2 | Rat hepatoma [HGPRT-] |
94101907 | C2-Rev 7 | Rat hepatoma [HGPRT-] |
89042701 | Fao | Rat hepatoma |
87031301 | H-4-II-E | Rat hepatoma Reuber H35 |
85061112 | H4-II-E-C3 | Rat liver hepatoma |
89102001 | H4S | Rat liver hepatoma |
94101905 | H5 | Rat hepatoma |
86062703 | Hep 3B | Human hepatocyte carcinoma |
85011430 | Hep G2 | Human Caucasian hepatocyte carcinoma |
92110305 | Hepa 1-6 | Mouse hepatoma |
95090613 | Hepa-1c1c7 | Mouse hepatoma |
94101908 | HF1 | Rat hepatoma Hybrid [H5xFao] |
94101909 | HF1-5 | Rat hepatoma Hybrid [H5xFao] |
93120108 | HTC | Rat hepatoma |
85061110 | HTC (BUdR) | Rat liver hepatoma |
1042712 | Huh-7D12 | Human hepatocellular carcinoma |
90021504 | MCA-RH 7777 | Rat Buffalo hepatoma |
96121721 | MH-22A | Mouse C3HA hepatocarcinoma |
90011902 | N1-S1 | Rat hepatoma |
91091816 | SK-HEP-1 | Human liver adenocarcinoma |