MilliporeSigma
Home3D Cell CultureCytoSoft® Modulus Plates for Cell Culture
CytoSoft® Modulus Plates for Cell Culture
What is the Elastic Modulus?
Young’s elastic modulus is the mechanical property that measures the stiffness of a material and is expressed in kilopascals of pressure (kPa). Cells interact with their local 3D microenvironments and reside in different matrix stiffnesses throughout the body, which varies based on tissue location or disease state.
Mechanotransduction is the processes through which cells sense and respond to mechanical stimuli. Cells convert the mechanical stimuli to biochemical signals that then elicit specific cellular responses. Mechanical force and regulatory signals are transmitted from the extracellular matrix (ECM) to the cell through focal adhesions, which are large macromolecular assemblies on the outside of the cell.
Elastic Modality Importance in Cell Culture
Human tissues and organs have low elastic modulus values that range from 0.2 to 64 kPa, whereas tissue cultureware has a much higher elastic modulus of 1x107 kPa. To create more relevant in vitro cell models, researchers have started to cultivate their cells on softer 3D hydrogels and substrates that more closely represent native tissue rigidities. For example, changing to a stiffer substrate alters the differentiation potential of human mesenchymal stem cells to favor bone formation over cartilage and adipose tissues1.
Native matrix stiffnesses of various in vivo tissues.
Figure 1. Native matrix stiffnesses of various in vivo tissues.
Cell TypeObservationReference
Mesenchymal Stem CellsMSCs on soft substrates (1 kPa) had a higher expression of chondrogenic marker collagen-II and adipogenic marker lipoprotein lipase, less spreading, fewer stress fibers and lower proliferation rates than on hard substrates (15 kPa).1
FibroblastsPDGF-induced chemotaxis and migration of pulmonary fibroblasts precultured on stiff substrates (25 kPa) were significantly higher than those of cells precultured on soft gels (2 kPa).2
Endothelial CellsEndothelial cell contraction-mediated neutrophil transmigration using an in vitro model of the vascular endothelium. Neutrophil transmigration increased with increasing substrate stiffness below the endothelium.3
Neural Stem CellsNSCs proliferated on substrates below 10 kPa and exhibited maximal proliferation on 3.5 kPa surfaces. Neuronal differentiation was favored on the softest surfaces with EY < 1 kPa. Oligodendrocyte differentiation was favored on stiffer scaffolds >7 kPa. Astrocyte differentiation was only observed on <1 and 3.5 kPa surfaces and represented less than 2% of the total cell population.4
Tumor CellsCCN1/CYR61 is highly regulated by stiffness in endothelial cells. Stiffness‐induced CCN1 activates β‐catenin nuclear translocation and signaling and that this contributes to upregulate N‐cadherin levels on the surface of the endothelium, increasing cancer cell metastasis.5
Cancer Stem CellsFibrotic rigidities found in pancreatic cancer tissues promote elements of EMT, including increases in vimentin expression, decreases in E-cadherin expression, nuclear localization of β-catenin, YAP and TAZ, changes in cell shape towards a mesenchymal phenotype and induction of chemoresistance.6
Mammary EpitheliumECM stiffness alone induces malignant phenotypes but that the effect is completely abrogated when accompanied by an increase in basement-membrane ligands.7
Hepatic Stellate CellsHigher matrix stiffness leads to transdifferentiation of hepatic stellate cells into fibrogenic, proliferative, and contractile myofibroblasts.8
CardiomyocytesInverse correlation between the material stiffness and the amplitude of contraction of the cardiac tissue constructs by changing the modulus of the matrix using different compositions of PEG and fibrinogen.9
Smooth Muscle CellsElastic modulus between 448 and 5804 Pa regulated SMC cytoskeletal assembly in 3D, with cells in stiff matrices having a slightly higher degree of F-actin bundling after prolonged culture.10
Table 1.Effects of matrix stiffness on cell behavior.
CytoSoft® Elastic Modulus Plates
An innovative tool to analyze the effect of matrix stiffness and rigidity on regulating cellular behavior, CytoSoft® elastic modulus plates are used to culture cells on substrates with various defined rigidities covering a broad physiological range (0.2kPA- 64kPa). On the bottom of each well, there is a thin layer of specially formulated biocompatible silicone, whose elastic modulus (rigidity) is carefully measured. The surfaces of the gels in CytoSoft® products are functionalized to form covalent bonds with amines on proteins. This chemical functionalization is stable and the reaction does not require a catalyst, facilitating the coating of the gel surfaces with matrix proteins and plating cells. For example, coating with an ECM protein, such as PureCol® (5006), is recommended before plating cells.
Fluorescent cell imaging of F-actin in HeLa cells using CytoSoft® imaging plates (8 kPA). Cell migration and adhesion can be analyzed by monitoring the expression of F-actin filamentous cytoskeleton protein.
Figure 2. Fluorescent cell imaging of F-actin in HeLa cells using CytoSoft® imaging plates (8 kPA).Cell migration and adhesion can be analyzed by monitoring the expression of F-actin filamentous cytoskeleton protein.
The silicone substrates of CytoSoft® plates are optically clear and have a low auto-florescence. The layer of silicone in each well is firmly bonded to the bottom of the well. Unlike hydrogels (such as polyacrylamide gels), silicone gels are not susceptible to hydrolysis, do not dry nor swell, are resilient and resistant to tearing or cracking, and their elastic moduli (rigidities) remain nearly unchanged during extended storage periods.
CytoSoft® products accommodate the harvesting of cells using enzymes such as trypsin and collagenase. There is no biochemical breakdown of the substrate during or after enzyme treatment, and there are no residuals of the substrate in the sample retrieved from a CytoSoft® plate.
For researchers who are unsure which CytoSoft® plate stiffness to use, we offer a Discovery Kit (5190). This kit contains various elastic moduli, including 0.2, 0.5, 2, 8, 16, 32 and 64 kPa in seven individual 6-well plates. Once the elastic modulus is determined, CytoSoft® Imaging plates can be selected. CytoSoft® Imaging plates have low autofluorescence and high optical clarity making them ideal for high-resolution imaging and live cell imaging.
Primary human dermal fibroblast matrix stiffness optimization.
Figure 3. Primary human dermal fibroblast matrix stiffness optimization.A elastic modulus of 8 kPA is an optimal matrix stiffness for dermal fibroblasts showing a reduction in F-actin stress fibers and increased cell adhesion (Vinculin) when compared with 0.2 or 64 kPa matrix stiffnesses.
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Park JS, Chu JS, Tsou AD, Diop R, Tang Z, Wang A, Li S. 2011. The effect of matrix stiffness on the differentiation of mesenchymal stem cells in response to TGF-?. Biomaterials. 32(16):3921-3930. https://doi.org/10.1016/j.biomaterials.2011.02.019
2.
Asano S, Ito S, Takahashi K, Furuya K, Kondo M, Sokabe M, Hasegawa Y. 2017. Matrix stiffness regulates migration of human lung fibroblasts. Physiol Rep. 5(9):e13281. https://doi.org/10.14814/phy2.13281
3.
Stroka KM, Aranda-Espinoza H. 2011. Endothelial cell substrate stiffness influences neutrophil transmigration via myosin light chain kinase-dependent cell contraction. 118(6):1632-1640. https://doi.org/10.1182/blood-2010-11-321125
4.
Leipzig ND, Shoichet MS. 2009. The effect of substrate stiffness on adult neural stem cell behavior. Biomaterials. 30(36):6867-6878. https://doi.org/10.1016/j.biomaterials.2009.09.002
5.
Reid SE, Kay EJ, Neilson LJ, Henze A, Serneels J, McGhee EJ, Dhayade S, Nixon C, Mackey JB, Santi A, et al. 2017. Tumor matrix stiffness promotes metastatic cancer cell interaction with the endothelium. EMBO J. 36(16):2373-2389. https://doi.org/10.15252/embj.201694912
6.
Rice AJ, Cortes E, Lachowski D, Cheung BCH, Karim SA, Morton JP, del Río Hernández A. 2017. Matrix stiffness induces epithelial?mesenchymal transition and promotes chemoresistance in pancreatic cancer cells. Oncogenesis. 6(7):e352-e352. https://doi.org/10.1038/oncsis.2017.54
7.
Chaudhuri O, Koshy ST, Branco da Cunha C, Shin J, Verbeke CS, Allison KH, Mooney DJ. 2014. Extracellular matrix stiffness and composition jointly regulate the induction of malignant phenotypes in mammary epithelium. Nature Mater. 13(10):970-978. https://doi.org/10.1038/nmat4009
8.
Wells RG. 2005. The Role of Matrix Stiffness in Hepatic Stellate Cell Activation and Liver Fibrosis. Journal of Clinical Gastroenterology. 39(Supplement 2):S158-S161. https://doi.org/10.1097/01.mcg.0000155516.02468.0f
9.
Shapira-Schweitzer K, Seliktar D. 2007. Matrix stiffness affects spontaneous contraction of cardiomyocytes cultured within a PEGylated fibrinogen biomaterial. Acta Biomaterialia. 3(1):33-41. https://doi.org/10.1016/j.actbio.2006.09.003
10.
Peyton SR, Kim PD, Ghajar CM, Seliktar D, Putnam AJ. 2008. The effects of matrix stiffness and RhoA on the phenotypic plasticity of smooth muscle cells in a 3-D biosynthetic hydrogel system. Biomaterials. 29(17):2597-2607. https://doi.org/10.1016/j.biomaterials.2008.02.005
11.
Wilson CL, Hayward SL, Kidambi S. Astrogliosis in a dish: substrate stiffness induces astrogliosis in primary rat astrocytes. RSC Adv.. 6(41):34447-34457. https://doi.org/10.1039/c5ra25916a
12.
Paul CD, Hruska A, Staunton JR, Burr HA, Daly KM, Kim J, Jiang N, Tanner K. 2019. Probing cellular response to topography in three dimensions. Biomaterials. 197101-118. https://doi.org/10.1016/j.biomaterials.2019.01.009
13.
Rossow L, Veitl S, Vorlová S, Wax JK, Kuhn AE, Maltzahn V, Upcin B, Karl F, Hoffmann H, Gätzner S, et al. 2018. LOX-catalyzed collagen stabilization is a proximal cause for intrinsic resistance to chemotherapy. Oncogene. 37(36):4921-4940. https://doi.org/10.1038/s41388-018-0320-2