MilliporeSigma
BRAND® - The Liquid Handling Company
BRAND® (BRANDTECH® in North America) is a leading Liquid Handling Company with more than 70 years of experience and a trusted partner and reference in the lab. Researchers and lab professionals around the world work with BRAND products in the life sciences, pharma, chemistry and process analytics. Our promise is to make laboratory work simple, easy, and efficient with high-quality products like the HandyStep® touch repetitive pipette, the Transferpette® S micropipette, the accu-jet® S pipette controller, the Dispensette® bottle-top dispenser or the BLAUBRAND® volumetric instruments.
We are part of the Brand Group which develops, manufactures and distributes high-quality and innovative laboratory equipment, vacuum pumps and systems with approx. 1,000 employees worldwide.
Products
Sort by Relevance
Showing 1-20 of 1998
Change View
Showing 1-20 of 1998
Compare
Product Number
Product Name
Product Description
Pricing
Brand® Liquid Handling
The Brand® liquid handling line, which includes the Dispensette® bottletop dispensers, Titrette® bottletop burettes, Transferpette® pipettes and HandyStep® repeating pipettes, sets the world standard for accuracy, ergonomic design, and reliability. In addition to pipette tips, filter tips, microcentrifuge tubes and PD-Tips, Brand® Life Science also provides a variety of other high performance products for PCR, storage, immunology and cell culture technology. The BRAND® Life Science Plastics line includes their innovative line of disposable UV-Cuvettes that allow photometric measurements down to 230 nm and fit nearly all spectrophotometers. The portfolio also includes PCR tubes, strips, plates and mats, microcentrifuge tubes, and 96-, 384-, and 1536-well plates.
Brand® Sample Preparation and Storage
Sample preparation and storage impose a wide variety of requirements on laboratory instruments. Brand® offers materials ranging from glass to specialty plastics, like PFA for trace analysis.
BLAUBRAND® Products
The BLAUBRAND® line from Brand® of volumetric instrument is individually calibrated for high precision. Statistical process control ensures with a certainty of at least 99.6 % (AQL = 0.4) compliance with predefined specifications. For the measurement of density, Brand® offers individually calibrated BLAUBRAND® density bottles, and fast, reliable hydrometers.