MilliporeSigma
HomeGas Chromatography (GC) MET-Biodiesel Capillary GC Column
MET-Biodiesel Capillary GC Column
MET-Biodiesel Capillary GC Column
The MET-Biodiesel was designed specifically for the determination of free and total glycerin in B100 biodiesel samples.
Features and benefits of this column include:
  • Good peak shape and resolution for all glyceride impurities of interest
  • Ability to separate glycerin in addition to mono-, di-, and triglycerides (as methyl esters) from the FAMEs
  • A maximum temperature of 380 °C (isothermal) and 430 °C (programmed), exceeding the temperature limitations specified in biodiesel methods ASTM® D6584 and EN 14105:2003
  • Metal was selected as the column material because it holds up better than fused silica under the method conditions, virtually eliminating column breakage
  • Integrated guard protects the analytical column from excess reagent and non-volatile compounds, extending column life with a leak-free connection
Chromatograms
The following chromatograms were generated using the MET-Biodiesel: (click to view)
Blank Containing Internal Standards Only on the MET-Biodiesel
ASTM® D6584 Calibration Standard on the MET-Biodiesel, Run 1 on a New Column
ASTM® D6584 Calibration Standard on the MET-Biodiesel, Run 100 on the Same Column
EN 14105 Monoglyceride Standard on the MET-Biodiesel
B100 Biodiesel Sample on the MET-Biodiesel
DescriptionCat. No.
MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µm with integrated 2 m x 0.53 mm I.D. guard28668-U
Ordering Information for GC Columns
Chromatograms
met-biodiesel-chomatogram
Blank Containing Internal Standards Only on the MET-Biodiesel
Peak IDs
1.Butanetriol (I.S.)
2.Tricaprin (I.S.)
Conditions
Column:MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U)
Oven:50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.)
Det.:FID, 380 °C
Carrier Gas:helium, 3.0 mL/min.
Injection:1 µL, cold on-column
Sample:Method blank consisting of Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane
met-biodiesel-astm
ASTM® D6584 Calibration Standard on the MET-Biodiesel, Run 1 on a New Column
Peak IDs
1.Glycerin
2.Butanetriol (I.S.)
3.Monoolein
4.Tricaprin (I.S.)
5.Diolein
6.Triolein
Conditions
Column:MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U)
Oven:50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.)
Det.:FID, 380 °C
Carrier Gas:helium, 3.0 mL/min.
Injection:1 µL, cold on-column
Sample:ASTM® D6584 Standard Solution 1 (44899-U) plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane
met-biodiesel
ASTM® D6584 Calibration Standard on the MET-Biodiesel, Run 100 on the Same Column
Peak IDs
1.Glycerin
2.Butanetriol (I.S.)
3.Monoolein
4.Tricaprin (I.S.)
5.Diolein
6.Triolein
Conditions
Column:MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U)
Oven:50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.)
Det.:FID, 380 °C
Carrier Gas:helium, 3.0 mL/min.
Injection:1 µL, cold on-column
Sample:ASTM® D6584 Standard Solution 1 (44899-U) plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane
met-biodiesel-en-standard
EN 14105 Monoglyceride Standard on the MET-Biodiesel
Peak IDs
1.Butanetriol (I.S.)
2.Monopalmitin (C16:0)
3.Monoolein (C18:1)
4.Monostearin (C18:0)
5.Tricaprin (I.S.)
Conditions
Column:MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U)
Oven:50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.)
Det.:FID, 380 °C
Carrier Gas:helium, 3.0 mL/min.
Injection:1 µL, cold on-column
Sample:EN 14105:2003 Monoglyceride Stock Solution (49446-U) plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane
met-biodiesel-b100
B100 Biodiesel Sample on the MET-Biodiesel
Peak IDs
1.Glycerin
2.Butanetriol (I.S.)
3.Monopalmitin (C16:0)
4.Monoolein (C18:1), Monolinolein (C18:2), Monolinolenin (C18:3)
5.Monostearin (C18:0)
6.Tricaprin (I.S.)
Conditions
Column:MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U)
Oven:50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.)
Det.:FID, 380 °C
Carrier Gas:helium, 3.0 mL/min.
Injection:1 µL, cold on-column
Sample:B100 Biodiesel plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane