Catalyst Screening & TLC-MS for Reaction Analysis
Sarah Trice, Thorsten vom Stein, Michael Schulz
Introduction
Catalyst screening is an important prerequisite in organic synthesis. KitAlysis™ High-Throughput Screening Kits showcase the most widely used catalytic methods of synthetic chemists, translated into an off-the-shelf screening system to quickly and efficiently identify or optimize suitable catalytic reaction conditions.
To evaluate the screening an analysis tool is needed. Thin layer chromatography (TLC) allows for a fast and cost-effective determination of the reaction progress by parallel analysis of the samples on one plate. This setup perfectly fits to the parallel synthesis utilizing KitAlysis™.
This application note displays the use of the KitAlysis™ High-Throughput Buchwald-Hartwig Amination Reaction Screening Kit for the optimization of the coupling of aryl bromide and diphenylamine combined with an evaluation of the result with TLC-MS analysis.
Figure 1.Schematic reaction showing the coupling of aryl bromide (1) and diphenylamine (2) to form biphenyl-4-yl-di-p-tolyl-amine (3), where the product has a molecular weight of 349.468 g/Mol.
Results and Discussion
The two reactants, aryl bromide and diphenylamine, were applied in addition to the samples. The samples were clearly visible under UV-light, therefore no derivatization was necessary. Figures 2a, 2b and 2c show the developed plate under white light, at 254 nm and at 366 nm.
Track | Samples / Catalysts KITALYSIS-CN | Concentration | Application volume | Rf of product and educts |
---|---|---|---|---|
1 | BrettPhos Pd G3 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
2 | CPhos Pd G3 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
3 | DavePhos Pd G3 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
4 | P(tBu3) Pd G2 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
5 | J009 Pd G3 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
6 | MorDalPhos Pd G3 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
7 | BINAP Pd G3 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
8 | RuPhos Pd G2 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
9 | SPhos Pd G2 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
10 | tBuXPhos Pd G3 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
11 | XantPhos Pd G3 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
12 | XPhos Pd G3 | KitAlysis™ sample filled up with 1 ml THF | 2.0 µL | 0.50 |
13 | amine educt | 1 mg/mL | 1.0 µL | 0.19 |
14 | Ar-Br educt | 1 mg/mL | 1.0 µL | 0.77 |
Figure 2a.Developed plate under white light
Figure 2b.Developed plate at 254 nm
Figure 2c.Developed plate at 366 nm
Confirmation of reaction product by mass spectrometry: Formation of ACN adduct via radical ionization
![Mass spectrum (ESI+) of product [M+ACN] Mass spectrum (ESI+) of product [M+ACN]](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/protocols/analytical-chemistry/thin-layer-chromatography/catalyst-screening-followed-by-tlc-ms/mass-spectrum/mass-spectrum.jpg)
Figure 3.Mass spectrum (ESI+) of product [M+ACN]
Biphenyl-4-yl-di-p-tolyl-amine (M=349.568 g/Mol)
Chromatography | Plate | HPTLC Silica gel 60 F254 MS-grade |
Sample preparation | Fill up the KiTAlysis™ samples with 1mL tetrahydrofuran (THF) Educts: Dissolve 1 mg each in 1 mL THF | |
Sample application | ATS 4 sample applicator (Camag), 6 mm bandwise | |
Application volume | 1.0 µl track 13-14 / 2.0 µL track 1-12 | |
Mobile phase | n-Heptane / dichloromethane 6/1 (v/v) + 0.1% trifluoroacetic acid | |
Migration distance | 5 cm | |
Migration time | 10-11 min | |
Extraction | Extraction equipment | Plate Express (Advion) |
Extraction solvent | Acetonitrile / water 95/5 (v/v) + 0.1 % formic acid | |
Extraction flow | 0.2 mL/min | |
Detection | Documentation equipment | TLC Visualizer (Camag) |
Wavelength | scan at white light, 254 nm, 366 nm | |
Staining | - | |
MS equipment | Single quadrupole mass spectrometer Expression CMS (Advion) | |
MS detection | ESI(+) mode (m/z 50 - 1200) |
Network error: Failed to fetch