MilliporeSigma
HomeSmall Molecule HPLCUSP HPLC Analysis of L-Methionine on Ascentis® Express OH5
USP HPLC Analysis of L-Methionine on Ascentis® Express OH5
USP HPLC Analysis of L-Methionine on Ascentis® Express OH5
Conditions
columnAscentis Express OH5 15 cm x 4.6 mm, 5 µm particles (50347-U)
column temp.40 °C
mobile phase[A] 0.005M Ammounium formate in Water (pH: 3.15 with Trifluoroacetic Acid) ), [B] Acetonitrile (30:70, A:B)
flow rate1.0 mL/min
pressure
914 psi (63 bar)
sample0.8 mg/mL water:acetonitrile (1:1)
injection3 µL
detectorAgilent 380-ELSD
Description
Analysis NoteThis USP application was generated by RTC on an Ascentis® Express OH5 column.
CategoriesAnalytical Chromatography, Amino Acids
Featured IndustryLife Science and Biopharma
suitabilityapplication for HPLC
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch