MilliporeSigma
US EPA Method 8041: GC Analysis of Phenols on Equity-5
US EPA Method 8041: GC Analysis of Phenols on Equity-5 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50
CONDITIONS
column
Equity-5, 30 m × 0.25 mm I.D., 0.25 μm (25312)
oven
40 °C, 8 °C/min to 300°C
inj. temp.
225 °C
detector
FID, 340 °C
carrier gas
helium, 25 cm/sec @ 60 °C
injection
1.0 μL 100:1 split
liner
split, cup design
sample
10 μg/mL each component (47899)
Description
Legal Information
null