GC Analysis of C2-C22 Free Fatty Acids on Nukol
Materials
Analytical column
Product No.
Description
Pricing
CONDITIONS
column
Nukol, 15 m × 0.53 mm I.D., 0.5 μm (25326)
oven
110 °C, 8 °C/min to 220 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 20 mL/min
injection
1 μL, direct
sample
25-400 μg/mL each analyte
Description
Legal Information
null