Product Name
Water suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Product Number
95283
Product Brand
SIAL
CAS Number
7732-18-5
Molecular Weight
18.02
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| DENSITY D20/4 | 0.997 - 0.999 |
| REFRACTIVE INDEX N20/D | 1.332 - 1.334 |
| TEST BY UV SPECTROSCOPY | A200 ≤0.05 |
| LUMINESCENCE | CORRESPONDS TO REQUIREMENTS |
| HPCE TEST | CORRESPONDS TO REQUIREMENTS |
| RECOMMENDED RETEST PERIOD | 24 MONTHS |