Product Name
Water for ultratrace analysis, ACS reagent
Product Number
14211
Product Brand
SIAL
CAS Number
7732-18-5
Molecular Weight
18.02
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| ALUMINIUM (ICP) | ≤ 0.5 ug/kg |
| BARIUM (ICP) | ≤ 0.5 ug/kg |
| BORON (ICP) | ≤ 0.0005 mg/kg |
| CALCIUM (ICP) | ≤ 0.5 ug/kg |
| CADMIUM (ICP) | ≤ 0.5 ug/kg |
| COBALT (ICP) | ≤ 0.5 ug/kg |
| CHROMIUM (ICP) | ≤ 0.5 ug/kg |
| COPPER (ICP) | ≤ 0.5 ug/kg |
| IRON (ICP) | ≤ 0.5 ug/kg |
| MERCURY (ICP) | ≤ 0.5 ug/kg |
| POTASSIUM (ICP) | ≤ 0.5 ug/kg |
| LITHIUM (ICP) | ≤ 0.5 ug/kg |
| MAGNESIUM (ICP) | ≤ 0.5 ug/kg |
| MANGANESE (ICP) | ≤ 0.5 ug/kg |
| MOLYBDENUM (ICP) | ≤ 0.5 ug/kg |
| SODIUM (ICP) | ≤ 0.5 ug/kg |
| LEAD (ICP) | ≤ 0.5 ug/kg |
| SELENIUM (ICP) | ≤ 0.005 mg/kg |
| SILICON (ICP) | ≤ 0.0005 mg/kg |
| TIN (ICP) | ≤ 0.5 ug/kg |
| STRONTIUM (ICP) | ≤ 0.5 ug/kg |
| TITANIUM (ICP) | ≤ 0.5 ug/kg |
| VANADIUM (ICP) | ≤ 0.5 ug/kg |
| ZINC (ICP) | ≤ 0.5 ug/kg |
| ZIRCONIUM (ICP) | ≤ 0.5 ug/kg |
| METAL TRACE ANALYSIS (ICP-MS) | CORRESPONDS TO REQUIREMENTS |
| RECOMMENDED RETEST PERIOD | 24 MONTHS |