50481
TrippNT
TrippNT® Vertical Glove Box and Kimwipe Holders
White PVC Vertical Glove Box and Kimwipe with Magnet Mount 5 3/4 in. x 31 1/4 in. x 4 1/4 in. (WHD)
Synonym(s):
Kimwipe holder
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
material
PVC
weight
2.63 lb
manufacturer/tradename
TrippNT
Related Categories
General description
TrippNT's White Large Kimwipe Holders provide a lean approach for cleaning laboratory and manufacturing equipment. Fits Kimwipes Kimberly Clark XL Wipers Part No. 34155, Accuwipe Part No. 29712 and Shur Wipe Wipers Part No. 29712. Made of white PVC with magnet mount. All TrippNT products are made in the USA and come with a 100% Total Replacement Lifetime Guarantee.
- Rectangular Tension Design with Bottom
- Magnet Backing for Easy Mounting
- White PVC Construction with Magnet Back
- Provides a Lean Approach to Cleaning Laboratory and Manufacturing Supplies
- Finish: Smooth, no rough or sharp edges
- Material: 1/4” White PVC
- Product Weight: 2.625 lbs
- Ships from: Kansas City, MO, USA
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.