Key Documents
WPC0500C
TriForest
TriForest Autoclavable Polycarbonate Wide Mouth Bottle / Centrifuge Flask
500mL, 72 per case
Synonym(s):
Autoclavable Bottles, Polycarbonate Bottles, Wide Mouth Bottles, Centrifuge Flasks
About This Item
PDF Icon
Shipped & Sold By TriForest
A Trusted Marketplace Seller
PDF Icon
Have a question?
feature
with cap
manufacturer/tradename
TriForest Enterprises
General description
TriForest polycarbonate (PC) labware is a safer alternative to glass. These bottles are engineered to withstand maximum speeds in all GS3 rotors, and they are Ideal for applications including ultra centrifugation or cuture of microbes, plants, algae, and organisms requiring large surface area to colume ratios.
  • Made with virgin polycarbonate resin, durable, chemically inert, non-leaching
  • Molded seal ring inside cap fits tightly against bottle neck
  • Tight-fitting closures (included) secure samples and prevent aerosol escape
  • Bottle and cap will both withstand reuse and multiple autoclave cycles
  • Manufactured in a Class 10K cleanroom
  • Sold by the case - 72 bottles per case
Choose from one of the most recent versions:
Certificates of Analysis (COA)