Key Documents
PGC2000M-C
TriForest
TriForest PETG Square Media / Packaging Bottle
2000mL, 12 per case
Synonym(s):
PETG Bottles, Media Packaging Bottles
About This Item
PDF Icon
Shipped & Sold By TriForest
A Trusted Marketplace Seller
PDF Icon
Have a question?
feature
with cap
manufacturer/tradename
TriForest Enterprises
General description
TriForest PETG Media / Packaging bottles are ergonomic, clear, and leakproof. Designed with automation in mind, these bottles are optimal for all filling, capping, and packaging operations. They are heavy duty and break-resistant, ideal for storage of valuable sera.
  • Thick walled and durable; shatter resistant
  • Excellent gas barrier properties; reduced permeability to CO2/O2
  • Molded-in graduations for approximate volume reference
  • Compliant with USP Class VI and FDA guidelines
  • Bottle and cap material are Certified Animal Derived Component Free
  • Manufactured in a Class 10K cleanroom
  • Sold by the case - 12 bottles per case