Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-1150

TriAltus™

TriAltus™ SUMO Protease

100 mg, Yeast protein overexpressed in E. coli with histidine (His) tag

Synonym(s):

SUMO, SUMO Protease, SUMO P, Ulp, Ulp1, Enzyme, SUMO Enzyme, Protease

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By TriAltus Bioscience
A Trusted Marketplace Seller
PDF Icon
Have a question?

General description

SUMO Protease can be used for cleaving both natural SUMO-modified proteins and recombinant SUMO fusion proteins. This powerful site-specific protease targets the tertiary structure of the Ubiquitin-like SUMO domain. It hydrolyzes the peptide bond in the x–Gly–Gly–x sequence after the di-glycine bond, ensuring precise cleavage without leaving any residual amino acids.


SPECIFICATIONS

PURITY | ~100% (SDS gel analysis)

ENZYME SOURCE | Yeast protein overexpressed in E. coli with histidine (His) tag

CLEAVAGE SITE | SUMO Protease cleaves after the C-terminal glycine at the end of the SUMO sequence. If the target protein is fused downstream of the SUMO C-terminus, SUMO Protease cleavage renders the native protein sequence without any additional amino acids.

CLEAVAGE EFFICIENCY | 1 µg protease will digest 2 mg protein with 99% efficiency at 4 °C in 40 minutes.

STORAGE/SHIPPING CONCENTRATION | ~7.5 mg/mL

SHIPPING CONDITIONS | We ship SUMO protease on dry ice in buffer (15 mM Tris (pH 8.0), 0.2 M NaCl, 50% glycerol).

RECOMMENDED STORAGE CONDITIONS | -80°C long term or -20°C for 2-3 months in shipping buffer

EXPIRATION | 6 months from receipt when stored as directed

SKU | 

1 mg: 30-1130

5 mg: 30-1135

10 mg: 30-1140

50 mg: 30-1146

100 mg: 30-1150


Additional processing information: This highly efficient SUMO protease is compatible with TriAltus™ CLiM protein affinity purification technology.

Legal Information

Use of TriAltus products falls under our limited license agreement and is for non-profit research use only. For commercial use, please contact us for licensing.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library