Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SPR-324E

StressMarq Biosciences Inc

StressMarq Alpha Synuclein Pre-formed Fibrils (Type 1)

100 µg, pack of 5

Synonym(s):

Alpha synuclein PFFs, Alpha synuclein aggregates, Non-A beta component of AD amyloid protein, NACP protein, SNCA protein, PARK1 protein, SYN protein, Parkinson disease familial 1 Protein

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
14.46
PDF Icon
Shipped & Sold By StressMarq Biosciences Inc.
A Trusted Marketplace Seller
PDF Icon
Have a question?

assay

95

concentration

2 mg/mL

application(s)

life science and biopharma

biological source

mouse

recombinant

E. coli

mol wt

predicted mol wt 14.46 kDa

pH

7.4

storage temp.

-80 °C

storage condition

avoid repeated freeze/thaw cycles

manufacturer/tradename

StressMarq Biosciences Inc

Related Categories

General description

Alpha Synuclein Pre-formed Fibrils (Type 1) are expressed in E. coli and Ion-exchange Purified. Certified >95% pure using SDS-PAGE analysis. Low endotoxin <5 EU/mL @ 2mg/mL. Validated for WB | SDS-PAGE | In vivo assay | In vitro assay. Monomer source is catalog# SPR-323.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library