Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA039836

Sigma-Aldrich

Anti-RIN3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Flj22439, Anti-Ras and rab interactor 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

AKKNLPTAPPRRRVSERVSLEDQSPGMAAEGDQLSLPPQGTSDGPEDTPRESTEQGQDTEVKASDPHSMPELPRTAKQPPVPPPRKKRISRQLASTLPA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIN3(79890)

General description

Ras and Rab interactor 3 (RIN3) is a novel Rab5-binding protein, encoded by the gene mapped to human chromosome 14q32. The encoded protein belongs to the RIN family of RAS effectors. RIN3 contains a SH2 domain, proline-rich domain, RH domain, vacuolar protein sorting-associated protein (Vps9) domain and ras-associating (RA) domain in the N- terminal region. RIN3 show tissue-specific expression and is highly expressed in mast cells.

Immunogen

Ras and Rab interactor 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-RIN3 antibody produced in rabbit has been used in immunohistochemistry.[1]
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Ras and Rab interactor 3 (RIN3) plays an essential role in vesicular trafficking by interacting with small GTPases such as Ras and Rab14. In addition, it also interacts with amphiphysin II and promotes receptor-mediated endocytosis. Mutations in the gene increases the risk of susceptibility to Paget′s disease of bone (PDB). The encoded protein acts as a guanine nucleotide exchange factor (GEF) for Rab 5 and Rab31. RIN3 plays an important role in basic mast cell functions and pathologies such as mast cell mediated chronic inflammation and mast cell hyperproliferation by regulating receptor tyrosine kinase (KIT) activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79570

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of RIN3 as a guanine nucleotide exchange factor for the Rab5 subfamily GTPase Rab31.
Kajiho H
The Journal of Biological Chemistry, 286(27), 24364-24373 (2011)
RIN3: a novel Rab5 GEF interacting with amphiphysin II involved in the early endocytic pathway.
Kajiho H
Journal of Cell Science, 116(Pt 20), 4159-4168 (2003)
Mahéva Vallet et al.
Human molecular genetics, 24(11), 3286-3295 (2015-02-24)
Paget's disease of bone (PDB) is a common disorder with a strong genetic component characterized by increased but disorganized bone remodelling. Previous genome-wide association studies identified a locus on chromosome 14q32 tagged by rs10498635 which was significantly associated with susceptibility
Genome-wide association identifies three new susceptibility loci for Paget's disease of bone.
Albagha OM
Nature Genetics, 43(7), 685-689 (2011)
RIN3 is a negative regulator of mast cell responses to SCF.
Janson C
PLoS ONE, 7(11), e49615-e49615 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service