Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA029143

Sigma-Aldrich

Anti-LTN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C21orf10, Anti-C21orf98, Anti-FLJ11053, Anti-KIAA0714, Anti-RNF160, Anti-Ring finger protein 160, Anti-ZNF294

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LMGVYIGSVMPNDSEWEKMRQSLPMQWLHRPLLEGRLSLNYECFKTDFKEQDIKTLPSHLCTSALLSKMVLIALRKETVL

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF160(26046)

General description

LTN1 (Listerin) is an E3 ubiquitin ligase, that is incorporated with ribosomes. It is of 180kDa and has an N terminus, HEAT/ARM (Huntingtin, elongation factor 3/Armadillo) repeats along with a C-terminal catalytic RING (really interesting new gene) domain. It is located on Chr21q22.11.

Immunogen

ring finger protein 160 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In yeast, LTN1 (Listerin) play an important role in quality control of nonstop proteins. It helps to mediate the ubiquitylation and subsequent degradation of translationally arrested nonstop proteins. Absence of Ltn1 in cells, show greater sensitivity to translation-specific drugs. In yeast, lack of Ltn1 leads to aggregation of toxic proteins, that may describe the embryonic lethality and neurodegenerative phenotypes of Ltn1/Listerin-mutant mice.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78851

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of a ribosome-associated E3 ubiquitin ligase in protein quality control.
Bengtson MH1and Joazeiro CA
Nature, 467(7314), 470-473 (2010)
Single-particle EM reveals extensive conformational variability of the Ltn1 E3 ligase.
Lyumkis D, et al.
Proceedings of the National Academy of Sciences of the USA, 110(5), 1702-1707 null
Comprehensive investigation of DNA methylation and gene expression in trisomy 21 placenta.
Lim JH, et al.
Placenta, 42, 17-24 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service