Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA021771

Sigma-Aldrich

Anti-CHRDL2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BNF-1, Anti-Breast tumor novel factor 1, Anti-Chordin-like protein 2, Anti-Chordin-related protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RVLVHTSVSPSPDNLRRFALEHEASDLVEIYLWKLVKGIFHLTQIKKVRKQDFQKEAQHFRLLAGPHEGHWNVFLAQTLELKVTASPDKVTKT

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHRDL2(25884)

General description

CHRDL2 (Chordin-like 2) is a novel chordin-related gene belonging to the chordin family of proteins. It encodes a putative extracellular matrix protein consistsing of a signal peptide and three cysteine-rich (CR) domains. In humans, it is expressed in several normal adult tissues, predominantly in liver, heart, prostate, testis, and ovary.

Immunogen

Chordin-like protein 2 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CHRDL2 (Chordin-like 2) may play a role in the cartilage regeneration, myoblast and osteoblast differentiation. It has been reported that CHRDL2 may negatively regulate the cartilage formation/regeneration from immature mesenchymal cells. CHRDL2 restricts BMPs activity by blocking their interaction with its type I BMP receptor ectodomain. Increased expression of CHRDL2 has been reported in several cancerous tissues such as breast, lung and colon cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74462

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anat Oren et al.
Gene, 331, 17-31 (2004-04-20)
Chordin-like cysteine-rich repeats (CRs) are conserved domains present in an expanding family of secreted proteins that associate with members of the TGF beta superfamily. In this study, we report the molecular cloning and characterization of CHL2 (chordin-like 2), a novel
Inmin Wu et al.
Gene, 311, 105-110 (2003-07-11)
In an effort to identify novel genes relevant to tumor angiogenesis, we compared the genes expressed in a matched pair composed of vascularized breast tumor and its adjacent normal tissue obtained from the same cancer patient. Using differential display, we
Naoki Nakayama et al.
Development (Cambridge, England), 131(1), 229-240 (2003-12-09)
We have identified a novel chordin-like protein, CHL2, which is structurally most homologous to CHL/neuralin/ventroptin. When injected into Xenopus embryos, CHL2 RNA induced a secondary axis. Recombinant CHL2 protein interacted directly with BMPs in a competitive manner to prevent binding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service