Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA020415

Sigma-Aldrich

Anti-EDIL3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DEL1, Anti-EGF-like repeats and discoidin I-like domains 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 1:100- 1:250
immunohistochemistry: 1:20- 1:50

immunogen sequence

NGGICTDLVANYSCECPGEFMGRNCQYKCSGPLGIEGGIISNQQITASSTHRALFGLQKWYPYYARLNKKGLINAWTAAE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EDIL3(10085)

General description

EGF-like repeats and discoidin I-like domains 3 (EDIL3) is a 52kDa protein secreted by endothelial cells. It has three epidermal growth factor (EGF)-like domains. EDIL3 is expressed in the brain, heart, kidney and small intestine.[1]

Immunogen

EGF-like repeats and discoidin I-like domains 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

EGF-like repeats and discoidin I-like domains 3 (EDIL3) interacts with integrin αvβ3 and takes part in vascular remodeling. It stimulates the adhesion of endothelial cells by interacting with integrin receptors. EDIL3 acts like a connection between phagocytes and cells targeted for apoptosis. It binds to phosphatidylserine (PS) and then acts as an opsonin to attach apoptotic cells to phagocytes. EDIL3 may be useful in antiangiogenic therapy for hepatocellular carcinoma. It has been shown that it stimulates tumor growth by increasing vascular formation.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75165.

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The RGD finger of Del-1 is a unique structural feature critical for integrin binding.
Schurpf T, et al.
Faseb Journal, 26(8), 3412-3420 (2012)
Xiaolong Zou et al.
Journal of biomedical science, 16, 33-33 (2009-03-19)
Developmentally regulated endothelial cell locus-1 (Del1) is an embryonic angiogenic factor expressed in early embryonic endothelial cells, but recently has been found to be expressed in some forms of cancers including colon and breast cancers, and melanoma, and human cancer
Jian-Cong Sun et al.
World journal of gastroenterology, 16(36), 4611-4615 (2010-09-22)
To determine the role of epidermal growth factor-like repeats and discoidin I-like domains 3 (EDIL3) in pathogenesis of hepatocellular carcinoma (HCC) by investigating the EDIL3 expression in HCC and its prognostic value for HCC. EDIL3 expression was detected in 101

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service