Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA018539

Sigma-Aldrich

Anti-MTL5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CXCDC2, Anti-metallothionein-like 5, testis-specific (tesmin)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

SSQELEDASCCSLKKDSNPMVICQLKGGTQMLCIDNSRTRELKALHLVPQYQDQNNYLQSDVPKPMTALVG

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MTL5(9633)

Immunogen

metallothionein-like 5, testis-specific (tesmin) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74485.

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tadashi Matsuura et al.
Journal of inorganic biochemistry, 88(2), 183-191 (2002-01-23)
Tesmin 60, a novel testis-specific gene, has been identified to have homology in plant and animal species, sharing a pair of cysteine-rich regions reported to be similar to metallothionein. The functional implications for these homologs, however, are not fully understood.
Tomas Eckschlager et al.
Current protein & peptide science, 10(4), 360-375 (2009-08-20)
Metallothioneins (MTs) are low molecular, cysteine-rich proteins that have naturally-occurring Zn(2+) in both clusters. They may serve as a reservoir of metals for synthesis of apoenzymes and zinc-finger transcription regulators. MTs are also involved with several important proteins e.g. p53
Christian Olesen et al.
Molecular reproduction and development, 67(1), 116-126 (2003-12-04)
Tesmin is a protein with homology to the metal-binding motif of the metallothionein protein family. Tesmin has been described as a testis-specific transcript, which starts to accumulate in 8-day-old mouse spermatocytes. Herein, a differential display comparing meiotic gene expression in
Shizuyo Sutou et al.
Biology of reproduction, 68(5), 1861-1869 (2003-02-28)
Tesmin is a testis-specific protein. Four mouse tesmin cDNAs so far reported encode a testis-specific, metallothionein-like, 30-kDa protein (tesmin-30). An antibody against tesmin-30, however, detected a protein of 60 kDa (tesmin-60) from the mouse testis. To resolve the relationship between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service