콘텐츠로 건너뛰기
Merck

HPA014485

Anti-FZD10 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

동의어(들):

Anti-CD350 antigen, Anti-Frizzled-10 precursor, Anti-Fz-10, Anti-FzE7, Anti-hFz10


조직 및 계약 가격을 보려면 로그인를 클릭합니다.

크기 선택

100 μL

₩924,259

₩924,259


구입 가능 여부는 고객센터에 문의하십시오.


제품정보 (DICE 배송 시 비용 별도)

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Human Protein Atlas Number:

다음으로 건너뛰기

기술 서비스
도움이 필요하신가요? 저희 숙련된 과학자 팀이 도와드리겠습니다.
도움 문의

제품 이름

Anti-FZD10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KTLQSWQQVCSRRLKKKSRRKPASVITSGGIYKKAQHPQKTHHGKYEIPAQSPTCV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Quality Level

Gene Information

human ... FZD10(11211)

유사한 품목 비교

전체 비교 보기

차이점 표시

1 of 4

이 품목
HPA043536HPA053970HPA045025
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Immunogen

frizzled class receptor 10

Other Notes

Corresponding Antigen APREST72738

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Frizzled-10 precursor (FZD10) in association with Wnt7b initiates canonical signaling. FZD10-Wnt7b combination enhances the β-catenin concentration in transfected cells. In lung airway epithelium, FZD10 might be an important FZD receptor to transfer Wnt7b signals. FZD10 enhances the phosphorylation of Dishevelled 2 (Dvl2), Dishevelled 3 (Dvl3) and triggers the Rac1-JNK (Jun amino-terminal kinases) signaling pathway leading to actin cytoskeleton structure rearrangement and increased anchorage-independent cell growth.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

General description

Frizzled-10 precursor (FZD10) synonym CD350 (cluster of differentiation 350) antigen is a seven transmembrane Wnt-signaling receptor. It is a member of frizzled family. FZD10 is expressed in a tissue specific pattern. It is found in airway epithelium, vascular smooth muscle and in lungs. It is mapped on chromosome 12q24.33.

적합한 제품을 찾을 수 없으신가요?  

당사의 제품 선택기 도구.을(를) 시도해 보세요.

저장 등급

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


가장 최신 버전 중 하나를 선택하세요:

시험 성적서(COA)

Lot/Batch Number

적합한 버전을 찾을 수 없으신가요?

특정 버전이 필요한 경우 로트 번호나 배치 번호로 특정 인증서를 찾을 수 있습니다.

이 제품을 이미 가지고 계십니까?

문서 라이브러리에서 최근에 구매한 제품에 대한 문서를 찾아보세요.

문서 라이브러리 방문

Molecular cloning of Frizzled-10, a novel member of the Frizzled gene family.
Koike J, et. al
Biochemical and Biophysical Research Communications, 262(1) (1999)
J Koike et al.
Biochemical and biophysical research communications, 262(1), 39-43 (1999-08-17)
The Frizzled genes encode WNT receptors. Frizzled-10 (FZD10), a novel member of the Frizzled gene family, has been cloned and characterized. Nucleotide sequence analysis showed that human FZD10 gene encodes a seven-transmembrane-receptor of 581 amino acids, with the N-terminal cysteine-rich
C Fukukawa et al.
Oncogene, 28(8), 1110-1120 (2009-01-13)
We previously reported that Frizzled homologue 10 (FZD10), a member of the Wnt signal receptor family, was highly and specifically upregulated in synovial sarcoma and played critical roles in its cell survival and growth. We here report a possible molecular

질문

후기

평점 값 없음

활성 필터

자사의 과학자팀은 생명 과학, 재료 과학, 화학 합성, 크로마토그래피, 분석 및 기타 많은 영역을 포함한 모든 과학 분야에 경험이 있습니다..

고객지원팀으로 연락바랍니다.