Skip to Content
MilliporeSigma

HPA014259

Anti-PTGDR2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD294, Anti-CRTH2, Anti-DP2, Anti-GPR44

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

RDTISRLDGRIMCYYNVLLLNPGPDRDATCNSRQAALAVSKF

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GPR44(11251)

Immunogen

Putative G-protein coupled receptor 44 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A E El-Shazly et al.
International immunopharmacology, 11(11), 1864-1870 (2011-08-13)
Prostaglandin D2 (PGD2) receptor CRTH2, is a pro-inflammatory molecule involved in eosinophil recruitment to the allergic airway. We investigated the expression of CRTH2 in eosinophil from allergic rhinitis patients (AR) and tested the modulatory role of several TH1 and TH2
Cecilia Lindskog et al.
Journal of proteomics, 75(9), 2611-2620 (2012-04-03)
Beta cell-specific surface targets are required for non-invasive monitoring of beta cell mass, which could be used for evaluation of new diabetes treatments as well as to help unravel pathogenic mechanisms underlying beta cell dysfunction. By antibody-based proteomics, we have
Kathleen R Bartemes et al.
The Journal of allergy and clinical immunology, 134(3), 671-678 (2014-08-31)
In mice, group 2 innate lymphoid cells (ILC2s) likely mediate helminth immunity, inflammation, and tissue repair and remodeling. However, the involvement of ILC2s in human diseases, such as asthma, is not well understood. The goals of this study were to