Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA013952

Sigma-Aldrich

Anti-CELSR2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cadherin EGF LAG seven-pass G-type receptor 2 precursor, Anti-Epidermal growth factor-like 2, Anti-Flamingo 1, Anti-Multiple epidermal growth factor-like domains 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Estimated to ship onMay 19, 2025



Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Estimated to ship onMay 19, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SATQDVHFTENLLRVGSALLDTANKRHWELIQQTEGGTAWLLQHYEAYASALAQNMRHTYLSPFTIVTPNIVISVVRLDKGNFAGAKLPRYEALRGEQPPDLETTVILPESVFRETPPVVRPAGPGEAQEPEELARRQRRHPELSQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CELSR2(1952)

General description

The gene CELSR2 (cadherin, EGF LAG seven-pass G-type receptor 2) encodes a member of the flamingo subfamily that comprises of nonclassic-type cadherins. The gene is mapped to human chromosome 1. It is also referred to as EGFL2 (epidermal growth factor (EGF)-like 2) and contains EGF-like repeats and cadherin repeat motifs. The encoded protein is 2923 amino acids in length and is 94% identical to mouse Flamingo protein. Its expression is strongest in the brain.

Immunogen

Cadherin EGF LAG seven-pass G-type receptor 2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The protein cadherin, EGF LAG seven-pass G-type receptor 2, encoded by the gene CELSR2 may play a role in neuronal cell-cell interaction. The exact function of this protein is yet to be determined.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72754

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The human homologue of flamingo, EGFL2, encodes a brain-expressed large cadherin-like protein with epidermal growth factor-like domains, and maps to chromosome 1p13.3-p21.1.
J B Vincent et al.
DNA research : an international journal for rapid publication of reports on genes and genomes, 7(3), 233-235 (2000-07-25)
Nilesh J Samani et al.
Journal of molecular medicine (Berlin, Germany), 86(11), 1233-1241 (2008-07-24)
Through genome-wide association studies, we have recently identified seven novel loci that confer a substantial increase in risk for coronary artery disease (CAD). Elucidating the mechanisms by which these loci affect CAD risk could have important clinical utility. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service