Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA009150

Sigma-Aldrich

Anti-NPHP3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CFAP31, Anti-FLJ30691, Anti-FLJ36696, Anti-KIAA2000, Anti-MKS7, Anti-NPH3, Anti-SLSN3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PEFAHSSIDVEGPFANVNRDDWDIAVASLLQVTPLFSHSLWSNTVRCYLIYTDETQPEMDLFLKDYSPKLKRMCETMGYFFHAVYFPIDVENQYLTVRKWEIEKS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPHP3(27031)

General description

NPHP3 (adolescent nephronophthisis 3) gene is localized to 3q21-22, and belongs to a group of proteins linked with nephronophthisis (NPHP). NPHP is a group of cystic kidney disorder, inherited in an autosomal recessive manner. NPHP3 protein is composed of 1330 amino acids, and in mice, it is expressed in retina, respiratory epithelium, neural tissues, biliary tract, kidney tubules, and liver.

Immunogen

Nephrocystin-3 recombinant protein epitope signature tag (PrEST)

Application

Anti-NPHP3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Null mutations in NPHP3 (adolescent nephronophthisis 3) are involved in the pathogenesis of renal-hepatic-pancreatic dysplasia (RHPD). It is an autosomal recessive disorder. In mice, homozygous missense mutation in this protein might be linked with polycystic kidney disease (pcy) phenotype. Mutations in this gene are linked with isolated NPHP cases and tapeto-retinal degeneration. It functions as an interacting partner of nephrocystin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71412

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaëlle Blandin et al.
Skeletal muscle, 3(1), 3-3 (2013-02-19)
The complexity of the skeletal muscle and the identification of numerous human disease-causing mutations in its constitutive proteins make it an interesting tissue for proteomic studies aimed at understanding functional relationships of interacting proteins in both health and diseases. We
Heike Olbrich et al.
Nature genetics, 34(4), 455-459 (2003-07-23)
Nephronophthisis (NPHP), a group of autosomal recessive cystic kidney disorders, is the most common genetic cause of progressive renal failure in children and young adults. NPHP may be associated with Leber congenital amaurosis, tapeto-retinal degeneration, cerebellar ataxia, cone-shaped epiphyses, congenital
Lawrence Copelovitch et al.
American journal of medical genetics. Part A, 161A(7), 1743-1749 (2013-05-21)
We report on five consecutive sibs three with fatal renal-hepatic-pancreatic dysplastic (RHPD) syndrome and two pregnancies ending in early abortion. Three of the fetuses reached term and two survived for 15 and 58 days. They had diffusely cystic kidneys with
Torunn Fiskerstrand et al.
The Journal of molecular diagnostics : JMD, 12(1), 125-131 (2009-12-17)
We have investigated a family where two siblings had a developmental disorder associated with polycystic dysplastic kidney disease that was incompatible with postnatal survival. Additional features observed were ductal plate malformation in the liver, dysplasia of the pancreas, and (in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service