Skip to Content
MilliporeSigma

HPA004153

Anti-SMOC1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SMOC-1 antibody produced in rabbit, Anti-SPARC-related modular calcium-binding protein 1 precursor antibody produced in rabbit, Anti-Secreted modular calcium-binding protein 1 antibody produced in rabbit

Sign In to View Organizational & Contract Pricing.

About This Item

Human Protein Atlas Number:
UNSPSC Code:
12352203

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: suitable

immunogen sequence

VMPSCESDARAKTTEADDPFKDRELPGCPEGKKMEFITSLLDALTTDMVQAINSAAPTGGGRFSEPDPSHTLEERVVHWYFSQLDSNSSNDINKREMKPFKRYVKKKAKPKKCARRFTDYCDLNK

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SMOC1(64093)

General description

SPARC related modular calcium binding 1 (SMOC1), an extracellular glycoproteins, belongs to the SPARC family.[1] It is localized and secreted in bone marrow derived mesenchymal stem cells (BMSCs) stimulated with osteogenic medium (OSM).[2] The protein is expressed in the developing optic stalk, ventral optic cup, and limbs of mouse embryos. It is composed of five domains: an N-terminal follistatin-like (FS) domain, two thyroglobulin-like (TY) domains similar to SMOC, and an extracellular calcium-binding (EC) domain.[1]

Immunogen

SPARC-related modular calcium-binding protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SMOC1 plays an essential role in ocular and limb development.[1] It is associated with the regulation of cell-matrix interaction by binding to many cell-surface receptors, the extracellular matrix, growth factors, and cytokines. It also functions in bone morphogenetic proteins (BMP) signaling cascade.[1] Mutation in SMOC1 causes a rare autosomal-recessive disorder, microphthalmia and limb anomalies (MLA).[3]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Other Notes

Corresponding Antigen APREST70561.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harold D Love et al.
PloS one, 4(12), e8384-e8384 (2009-12-23)
Benign prostatic hyperplasia (BPH) and prostate carcinoma (CaP) are linked to aging and the presence of androgens, suggesting that androgen regulated genes play a major role in these common diseases. Androgen regulation of prostate growth and development depends on the
Ippei Okada et al.
American journal of human genetics, 88(1), 30-41 (2011-01-05)
Microphthalmia with limb anomalies (MLA) is a rare autosomal-recessive disorder, presenting with anophthalmia or microphthalmia and hand and/or foot malformation. We mapped the MLA locus to 14q24 and successfully identified three homozygous (one nonsense and two splice site) mutations in
Young-Ae Choi et al.
Journal of proteome research, 9(6), 2946-2956 (2010-04-03)
Extracellular matrix proteins have been implicated in the regulation of osteoblast differentiation of bone marrow derived mesenchymal stem cells (BMSCs) through paracrine or autocrine mechanisms. In the current study, we analyzed the secretory protein profiles of BMSCs grown in osteogenic
Hana Abouzeid et al.
American journal of human genetics, 88(1), 92-98 (2011-01-05)
Waardenburg anophthalmia syndrome, also known as microphthalmia with limb anomalies, ophthalmoacromelic syndrome, and anophthalmia-syndactyly, is a rare autosomal-recessive developmental disorder that has been mapped to 10p11.23. Here we show that this disease is heterogeneous by reporting on a consanguineous family, not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service