Skip to Content
MilliporeSigma

HPA003299

Anti-ASB8 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ASB-8, Anti-Ankyrin repeat and SOCS box protein 8


Sign In to View Organizational & Contract Pricing.

Select a Size

100 μL

$615.00

$615.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Human Protein Atlas Number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Anti-ASB8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

QSIQSKYSLSERLIRTIAAIRSFPHDNVEDLIRGGADVNCTHGTLKPLHCACMVSDADCVELLLEKGAEVNALDGYNRTALHYAAEKDEACVEVLLEYGANPNALDGNRDTPLHWAAFKNNAECVRALLESGASVNALDYNN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Quality Level

Gene Information

human ... ASB8(140461)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA003300HPA055240WH0140462M1
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous solution

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ASB8 (ankyrin repeat and SOCS box containing 8) gene encodes a protein belonging to the ankyrin repeat and SOCS (suppressor of cytokine signaling) box containing protein family. It contains four Ankyrin repeats and one SOCS box. The gene contains four exons interspaced by three introns is mapped to chromosome 12q13. It is predominantly expressed in skeletal muscle and at varied levels in heart, brain, placenta, liver, kidney, and pancreas. It may be involved in the development of lung cancer. It may be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex that functions in ubiquitination and proteasome-mediated degradation of target proteins.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Immunogen

Ankyrin repeat and SOCS box protein 8 recombinant protein epitope signature tag (PrEST)

Other Notes

Corresponding Antigen APREST86450

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Aaen Andresen et al.
The Journal of biological chemistry, 289(4), 2043-2054 (2013-12-18)
The ankyrin and SOCS (suppressor of cytokine signaling) box (ASB) family of proteins function as the substrate recognition subunit in a subset of Elongin-Cullin-SOCS (ECS) E3 ubiquitin ligases. Despite counting 18 members in humans, the identity of the physiological targets

Related Content

Prestige Antibodies Immunofluorescence Procedure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service