Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0287

Sigma-Aldrich

Hongotoxin-1

>98% (HPLC), lyophilized powder, recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

recombinant

expressed in E. coli

assay

>98% (HPLC)

form

lyophilized powder

mol wt

calculated mol wt 4226 Da

solubility

aqueous buffer: soluble

storage temp.

−20°C

Amino Acid Sequence

Thr-Val-Ile-Asp-Val-Lys-Cys-Thr-Ser-Pro-Lys-Gln-Cys-Leu-Pro-Pro-Cys-Lys-Ala-Gln-Phe-Gly-Ile-Arg-Ala-Gly-Ala-Lys-Cys-Met-Asn-Gly-Lys-Cys-Lys-Cys-Tyr-Pro-His

Biochem/physiol Actions

Blocks cloned and heterologously expressed Kv1.1, Kv1.2, Kv1.3 and Kv1.6 voltage-gated potassium channels.

Other Notes

Originally isolated from Centruroides limbatus.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service