Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

CWA-1024

Sigma-Aldrich

Anti-Human CD35 (E11) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
E11, monoclonal
application:
FACS
species reactivity:
human
technique(s):
flow cytometry: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

E11, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 223.66 kDa

species reactivity

human

species reactivity (predicted by homology)

nonhuman primates

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG1

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CR1(1378)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone E11 is a mouse monoclonal antibody that detects human Complement receptor type 1 (CD35).

Immunogen

Acute monocytic leukemia cells and normal human blood monocytes.

Application

Quality Control Testing:

Evaluated by Flow Cytometry in Human peripheral blood mononuclear cells (PBMC).

Flow Cytometry Analysis: Staining of one million Human peripheral blood mononuclear cells (PBMC) was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1024, Anti-CD35 Antibody (E11) ColorWheel® Mouse mAb and Cat. No. CWD-PE ColorWheel® activated Phycoerythrin (PE) Dye or an equivalent amount of PE-conjugated Mouse IgG1 isotype control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Complement receptor type 1 (UniProt: P17927; also known as C3b/C4b receptor, CD35) is encoded by the CR1 (also known as C3BR) gene (Gene ID: 1378) in human. CD35 is a single-pass type I membrane glycoprotein of the receptors of complement activation (RCA) family. It is synthesized with a signal peptide (aa 1-42), which is subsequently cleaved off to generate the mature form that contains a long extracellular domain (aa 42-1971) , a transmembrane domain (aa 1972-1996), and a cytoplasmic domain (aa 1997-2039). It contains 30 Sushi domains. Its expression has been reported on erythrocytes, a subset of T-cells, mature B cells, follicular dendritic cells, monocytes, and granulocytes. It serves as a membrane immune adherence receptor that plays a critical role in the capture and clearance of complement -opsonized pathogens by erythrocytes and monocytes/macrophages. Its ligands include C3b, iC3b, and C4b fragments of C3 and C4 proteins. CD35 also acts as an inhibitor of spontaneous complement activation by impairing the formation and function of the alternative and classical pathways C3/C5 convertases, and by serving as a cofactor for the cleavage of C3b to iC3b, C3c, and C3d, C3g, and C4b, C4c, and C4d by factor I. It also plays a role in immune regulation by contributing, upon ligand binding, to the generation of regulatory T cells from activated helper T cells and inn maintaining a balance between normal and pathological B-cell activation. (Ref.: Mácsik-Valent, B., et al. (2019). Front. Immunol. 10; 1493; Kremlitzka, M., et al. (2013). Int. Immunol. 25(1); 25-33; Schifferli, JA., et al. (1988). J. Immunol. 140(3); 899-904).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service