Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

CWA-1019

Sigma-Aldrich

Anti-Human CD3 (UCHT1) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

UCHT1, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 23.15 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone UCHT1 is a mouse monoclonal antibody that detects human T-cell surface glycoprotein CD3.

Immunogen

Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor.

Application

Quality Control Testing:

Evaluated by Flow Cytometry in Human peripheral blood mononuclear cells (PBMC).

Flow Cytometry Analysis (FC): Staining of one million Human peripheral blood mononuclear cells (PBMC) was performed using 5 μL of 1:1 mixture of Cat. No. CWA-1019, Anti-Human CD3 (UCHT1) Colorwheel® Dye-Ready mAb and Cat. No. CWD-PE Colorwheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of a PE-conjugated Mouse IgG1 isotype control.

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

T-cell surface glycoprotein CD3 epsilon chain (UniProt: P07766; also known as T-cell surface antigen T3/Leu-4 epsilon chain, CD3e) is encoded by the CD3E (also known as T3E) gene (Gene ID: 916) in human. CD3 is a single-pass type I membrane protein that is a part of the TCR-CD3 complex present on T-cells and plays an essential role in adaptive immune response. The TCR CD3 complex is formed through noncovalent association of TCR with a CD3 signalling apparatus consisting of the , , and subunits. It is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 23-126), a transmembrane protein (aa 127-152), and a cytoplasmic domain (aa 153-207). The CD3 complex performs two major functions. Firstly, CD3 chains are required for intracellular signaling where tyrosines in its immunoreceptor tyrosine-based activation motif (ITAM) domain (aa 178-205) are phosphorylated by Lck. Secondly, the CD3 complex is involved in TCR surface expression. In the endoplasmic reticulum, the TCR heterodimer physically associate with the CD3 complex prior to moving to the Golgi for glycosylation and subsequently translocating to the T-cell surface. Mutations in CD3E gene are known to cause immunodeficiency 18 (IMD18) that is characterized by recurrent infections in infancy or early childhood with variable severity. (Ref.: Dong, D., et al. (2019). Nature. 573(7775); 546-552).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service