Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9118

Sigma-Aldrich

Anti-CHIP (N-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CHIP, carboxy terminus of Hsp70p-interacting protein, Anti-HSPABP2, Anti-NY-CO-7, Anti-STUB1, STIP1 homology and U-box containing protein 1, Anti-UBOX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~35 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cells lysate and mouse brain extract (S2 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Application

Anti-CHIP (N-terminal) antibody produced in rabbit is suitable for immunoblotting at a working concentration of 1-2 μg/mL using HEK-293T cells lysate and mouse brain extract (S2 fraction). It was used for western blotting of the eluted immunoprecipitates of PHF-1 antibody or anti-FLAG M2 in a study.

Biochem/physiol Actions

CHIP is a dual-function chaperone/E3 ubiquitin ligase that plays a crucial role in regulating protein quality control at multiple levels. It is comprised of a N-terminal tetratricopeptide repeat (TPR), a U-box at its C-terminus and a highly charged internal region. It ubiquitinates Hsp70, a cytosolic chaperone and enhances refolding of stress damaged proteins. Additionally, it has E3 ubiquitin ligase activity and triggers proteasome degradation of irreversibly damaged proteins to prevent cellular toxicity. The ubiquitination, aggregation and degradation of several proteins involved in neurodegenerative disorders including tau, huntingtin, Cu/Zn SOD1, ataxin-1, and α-synuclein are regulated by CHIP and Hsp70. The aggregation and toxicity of polyglutamine (polyQ) expanded proteins is suppressed by CHIP. It also regulates stress-dependent apoptosis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonard Petrucelli et al.
Human molecular genetics, 13(7), 703-714 (2004-02-14)
Molecular chaperones, ubiquitin ligases and proteasome impairment have been implicated in several neurodegenerative diseases, including Alzheimer's and Parkinson's disease, which are characterized by accumulation of abnormal protein aggregates (e.g. tau and alpha-synuclein respectively). Here we report that CHIP, an ubiquitin
Jae Ryoung Hwang et al.
Cell stress & chaperones, 10(2), 147-156 (2005-07-26)
Apoptosis signal-regulating kinase 1 (ASK1) is a mitogen-activated protein kinase kinase kinase (MAPKKK) that is regulated under conditions of cellular stress. ASK1 phosphorylates c-Jun N-terminal kinase (JNK) and elicits an apoptotic response. ASK1 activity is regulated at multiple levels, 1
Victor M Miller et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 25(40), 9152-9161 (2005-10-07)
Huntington's disease (HD) and other polyglutamine (polyQ) neurodegenerative diseases are characterized by neuronal accumulation of the disease protein, suggesting that the cellular ability to handle abnormal proteins is compromised. As both a cochaperone and ubiquitin ligase, the C-terminal Hsp70 (heat
Daniel C Carrettiero et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(7), 2151-2161 (2009-02-21)
Tau inclusions are a prominent feature of many neurodegenerative diseases including Alzheimer's disease. Their accumulation in neurons as ubiquitinated filaments suggests a failure in the degradation limb of the Tau pathway. The components of a Tau protein triage system consisting
Niubys Cayado-Gutiérrez et al.
Cell stress & chaperones, 18(2), 243-249 (2012-08-22)
Hsp27 (HSPB1) is usually overexpressed in breast cancers affecting the disease outcome and the sensitivity of tumors to chemotherapy and radiotherapy. Hsp27 interacts with other proteins such as β-catenin, histone deacetylase HDAC6, transcription factor STAT2 and procaspase-3. Phosphatase and tensin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service