Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

OM-200050X

OMICRON™

Omicron™ Hydrophobic PTFE Vent Filter

Pore Size 0.45 μm, diam. 50 mm, STERILE, ¼-½in. hose barb, 10/Pack

Synonym(s):

PTFE filter, vent filter, vacuum line filter

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By Omicron Scientific
A Trusted Marketplace Seller
PDF Icon
Have a question?

General description

OMICRON™ Air Vent Filters STERILE

Media - PTFE (Polytetrafluoro ethylene)

Hydrophobic

Technical Specification
  • Sterilization: ETO
  • Max. Temperature: 60°C (140°F)
  • Bubble Point: >24 psi in 70% IPA
  • Air Flow Rate: 10 NLPM at 5 psi
  • Stepped Hose Barbs, (¼-½ in) diameter. Connections have internal taper design to accept male slip luer.
  • Effective Filtration Area: 16.2cm²
  • Housing Burst at 25°C: 75 psi (5.17 bar)
Applications
  • Pharmaceutical vents
  • Applicable for small volume venting & degassing venting bioreactors
  • Fermentation tanks & carboys sterile gas purging of culture vessel
  • Laboratory: clean or sterile: gases, slovents, reagents, drying gases
  • Electronics: photoresists, solvents, gases for research
Benefits of venting
  • Contamination control
  • Higher throughput, increased flow rates
  • High permeability rates
  • Increased systems suitability

(Similar to DS0222-0045)

Legal Information

OMICRON™ is a trademark of Omicron Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library